Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00432399_m1
          See other CD36 GE Assays ›
          Gene Symbol:
          Cd36
          Gene Name:
          CD36 antigen
          Gene Aliases:
          FAT, GPIV, Scarb3
          Chromosome Location:
          Chr.5: 17781690 - 17888959 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00432399_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd36
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159555.1 NP_001153027.1 4 - 5 739 125
          NM_001159556.1 NP_001153028.1 4 - 5 599 125
          NM_001159557.1 NP_001153029.1 5 - 6 650 125
          NM_001159558.1 NP_001153030.1 5 - 6 766 125
          NM_007643.4 NP_031669.3 4 - 5 742 125
          GenBank mRNA AK052825.1 - 4 - 5 543 125
          AK128908.1 - 5 - 6 543 125
          AK150648.1 - 4 - 5 563 125
          AK151355.1 - 4 - 5 578 125
          AK152904.1 - 2 - 3 249 125
          AK172162.1 - 5 - 6 588 125
          BC010262.1 - 4 - 5 551 3481681 125
          L23108.1 - 4 - 5 560 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001992.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd36

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          long-chain fatty acid metabolic process
          positive regulation of cytokine production
          positive regulation of cell-matrix adhesion
          pattern recognition receptor signaling pathway
          negative regulation of systemic arterial blood pressure
          fatty acid metabolic process
          triglyceride metabolic process
          transport
          receptor-mediated endocytosis
          phagocytosis, recognition
          cell adhesion
          signal transduction
          cell surface receptor signaling pathway
          nitric oxide mediated signal transduction
          response to mechanical stimulus
          negative regulation of gene expression
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of cholesterol storage
          fatty acid transport
          long-chain fatty acid transport
          long-chain fatty acid import across plasma membrane
          negative regulation of angiogenesis
          fatty acid oxidation
          lipid storage
          cGMP-mediated signaling
          positive regulation of blood coagulation
          cholesterol transport
          positive regulation of interleukin-12 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          plasma lipoprotein particle clearance
          low-density lipoprotein particle clearance
          lipoprotein transport
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          apoptotic cell clearance
          positive regulation of MAPK cascade
          regulation of transcription from RNA polymerase II promoter in response to oxidative stress
          long-chain fatty acid import into cell
          positive regulation of peptidyl-tyrosine phosphorylation
          defense response to Gram-positive bacterium
          low-density lipoprotein particle mediated signaling
          positive regulation of phagocytosis, engulfment
          positive regulation of macrophage cytokine production
          cellular response to bacterial lipopeptide
          cellular response to lipopolysaccharide
          cellular response to lipoteichoic acid
          cellular response to hydroperoxide
          regulation of removal of superoxide radicals
          positive regulation of blood microparticle formation
          positive regulation of reactive oxygen species metabolic process
          low-density lipoprotein particle receptor activity
          protein binding
          high-density lipoprotein particle binding
          lipid binding
          low-density lipoprotein particle binding
          transforming growth factor beta binding
          thrombospondin receptor activity
          oleic acid binding
          lipoteichoic acid immune receptor activity
          lipoprotein particle binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500018130.mm.1 TC0500001992.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018130.mm.1 NM_001159555.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018130.mm.1 NM_001159555.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018130.mm.1 NM_001159556.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018130.mm.1 NM_001159556.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018130.mm.1 NM_001159557.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018130.mm.1 NM_001159557.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018130.mm.1 NM_001159558.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018130.mm.1 NM_001159558.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018130.mm.1 NM_007643.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018130.mm.1 NM_007643.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018131.mm.1 TC0500001992.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018131.mm.1 NM_001159555.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018131.mm.1 NM_001159555.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018131.mm.1 NM_001159556.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018131.mm.1 NM_001159556.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018131.mm.1 NM_001159557.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018131.mm.1 NM_001159557.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018131.mm.1 NM_001159558.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018131.mm.1 NM_001159558.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018131.mm.1 NM_007643.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018131.mm.1 NM_007643.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0