Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433145_g1
          See other ESRRA GE Assays ›
          Gene Symbol:
          Esrra
          Gene Name:
          estrogen related receptor, alpha
          Gene Aliases:
          ERRalpha, Err1, Estrra, Nr3b1
          Chromosome Location:
          Chr.19: 6910977 - 6922048 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433145_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Esrra
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007953.2 NP_031979.2 6 - 7 1206 70
          GenBank mRNA AK156371.1 - 6 - 7 1182 70
          BC034165.1 - 6 - 7 1149 70
          BC039774.1 - 6 - 7 1218 70
          BC138586.1 - 6 - 7 1166 8861606 70
          BC171958.1 - 6 - 7 1166 9054244 70
          U85259.1 - 6 - 7 1170 70

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Esrra

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          regulation of ossification
          regulation of cell population proliferation
          steroid hormone mediated signaling pathway
          regulation of osteoblast differentiation
          regulation of osteoclast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          cartilage development
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity
          steroid hormone receptor activity
          steroid binding
          zinc ion binding
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936680 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936679 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936683 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936678 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936681 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954306 U85259.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954306 BC039774.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954306 BC052930.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954306 AK156371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954306 BC034165.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 937376 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 730808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765287 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245691 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935513 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 19257 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145671 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144711 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954306 U85259.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954306 BC039774.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954306 BC052930.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954306 AK156371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954306 BC034165.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P248580 U85259.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P248580 BC039774.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P248580 BC052930.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P248580 AK156371.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P248580 BC034165.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P198949 AK046588.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936669 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144711 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145671 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09097315 - -
          Whole Mouse Genome G4122A A_52_P198949 AK046588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P248580 U85259.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P248580 BC039774.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P248580 BC052930.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P248580 AK156371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P248580 BC034165.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108080 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145671 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144711 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14340223 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936681 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936683 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936680 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936678 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14248017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14262294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14421463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14216947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14262303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14468205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14468206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936648 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465487 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465489 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465488 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465492 - -
          Affymetrix HT MG-430 PM Array Plate 1460652_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1442864_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465485 - -
          Mouse Genome 430 2.0 Array 1442864_PM_at - -
          Mouse Genome 430 2.0 Array 1460652_PM_at - -
          Mouse Genome 430 2.0 Array 1442864_at - -
          Mouse Genome 430 2.0 Array 1460652_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26379_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26379_at - -
          Mouse Expression Set 430 1460652_PM_at - -
          Mouse Expression Set 430 1460652_at - -
          Mouse Expression Set 430 1442864_at - -
          Mouse Expression Set 430 1442864_PM_at - -
          Murine Genome U74v2 Set 103964_at - -
          Murine Genome U74v2 Set 102145_PM_f_at - -
          Murine Genome U74v2 Set 103964_PM_at - -
          Murine Genome U74v2 Set 102145_f_at - -
          Mouse Genome 430A 2.0 Array 1460652_PM_at - -
          Mouse Genome 430A 2.0 Array 1460652_at - -
          Affymetrix HT Mouse Genome 430A Array 1460652_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460652_at - -
          Affymetrix HT Mouse Genome 430B Array 1442864_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442864_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6550242 - -
          Illumina mouse-6 v1.0 expression beadchip 6550242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550242 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2766521 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2766521 - -
          mouse-6 v1.1 (Illumina) scl52911.9_189-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52911.9_189-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2972292 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2972292 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2766521 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl52911.9_189-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline