Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433918_g1
          See other CFB GE Assays ›
          Gene Symbol:
          Cfb
          Gene Name:
          complement factor B
          Gene Aliases:
          AI195813, AI255840, B, Bf, C2, Fb, H2-Bf
          Chromosome Location:
          Chr.17: 34856374 - 34862514 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433918_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cfb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001142706.1 NP_001136178.1 11 - 12 1905 62
          NM_008198.2 NP_032224.2 11 - 12 1905 62
          GenBank mRNA AK098094.1 - 11 - 12 1565 62
          AK149531.1 - 11 - 12 1538 62
          AK149573.1 - 11 - 12 1564 62
          AK151220.1 - 5 - 6 524 62
          AK151618.1 - 5 - 6 524 62
          AK151974.1 - 5 - 6 524 62
          BC005451.1 - 11 - 12 1539 3591669 62
          K01496.1 - 6 - 7 645 62
          K01498.1 - 1 - 2 69 62
          M57890.1 - 11 - 12 1608 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002816.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cfb

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          proteolysis
          complement activation
          complement activation, alternative pathway
          cell population proliferation
          innate immune response
          response to thyroid hormone
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413866 BP766400.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413866 K01496.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413866 M57890.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413866 AK151618.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413866 AK151974.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413866 BP766400.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413866 K01496.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413866 M57890.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413866 AK151618.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413866 AK151974.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359279 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345475 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 809727 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413866 BP766400.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413866 K01496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413866 M57890.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413866 AK151618.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413866 AK151974.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413866 BP766400.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413866 K01496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413866 M57890.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413866 AK151618.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413866 AK151974.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413866 BP766400.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413866 K01496.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413866 M57890.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413866 AK151618.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413866 AK151974.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413866 BP766400.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413866 K01496.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413866 M57890.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413866 AK151618.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413866 AK151974.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156201 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08766658 - -
          Whole Mouse Genome G4122A A_51_P413866 BP766400.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413866 K01496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413866 M57890.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413866 AK151618.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413866 AK151974.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413866 BP766400.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413866 K01496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413866 M57890.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413866 AK151618.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413866 AK151974.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138911 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13194686 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13194692 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13194679 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13194700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13404634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13194679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13404635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13515804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13515806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13341911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13515807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13341905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13404654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13194682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31156189 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450334 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450336 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450328 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450327 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450331 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450342 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450332 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450333 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450326 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450330 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450329 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450338 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450335 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450337 - -
          Affymetrix HT MG-430 PM Array Plate 1417314_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10450325 - -
          Mouse Genome 430 2.0 Array 1417314_PM_at - -
          Mouse Genome 430 2.0 Array 1417314_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14962_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14962_at - -
          Affymetrix Mouse Clariom S Array PSR1700018226.mm.1 TC1700002816.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018226.mm.1 NM_001142706.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018226.mm.1 NM_001142706.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018226.mm.1 NM_008198.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018226.mm.1 NM_008198.2 Refseq
          Mouse Expression Set 430 1417314_at - -
          Mouse Expression Set 430 1417314_PM_at - -
          Murine Genome U74v2 Set 100609_PM_at - -
          Murine Genome U74v2 Set 100609_at - -
          Murine Genome U74v2 Set 161365_r_at - -
          Murine Genome U74v2 Set 161365_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1417314_at - -
          Mouse Genome 430A 2.0 Array 1417314_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417314_at - -
          Affymetrix HT Mouse Genome 430A Array 1417314_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 730577 - -
          Illumina mouse-6 v1.0 expression beadchip 730577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000730577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230853 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230853 - -
          mouse-6 v1.1 (Illumina) scl50010.18.1_105-S - -
          Illumina Sentrix mouse-6.v1 bead array 730577 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50010.18.1_105-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 730577 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230853 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230853 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50010.18.1_105-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0