Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433925_g1
          See other SLC39A7 GE Assays ›
          Gene Symbol:
          Slc39a7
          Gene Name:
          solute carrier family 39 (zinc transporter), member 7
          Gene Aliases:
          AA408174, AI117660, AL024048, H-2Ke4, H2-Ke4, Ke-4, Ke4, Ring5, Zip7
          Chromosome Location:
          Chr.17: 34028266 - 34031690 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433925_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc39a7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077709.1 NP_001071177.1 2 - 3 581 64
          NM_008202.2 NP_032228.2 1 - 2 866 64
          GenBank mRNA AK159968.1 - 1 - 2 742 64
          BC115426.2 - 1 - 2 720 40093613 64
          BC115427.1 - 1 - 2 720 40093614 64
          KJ650218.1 - 1 - 2 434 64
          M32010.1 - 1 - 2 734 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002811.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc39a7

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          zinc ion transport
          metal ion transport
          transmembrane transport
          protein binding
          metal ion transmembrane transporter activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700017683.mm.1 TC1700002811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017683.mm.1 NM_001077709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017683.mm.1 NM_001077709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017683.mm.1 NM_008202.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017683.mm.1 NM_008202.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017685.mm.1 TC1700002811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017685.mm.1 NM_001077709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017685.mm.1 NM_001077709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017685.mm.1 NM_008202.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017685.mm.1 NM_008202.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017686.mm.1 TC1700002811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017686.mm.1 NM_001077709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017686.mm.1 NM_001077709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017686.mm.1 NM_008202.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017686.mm.1 NM_008202.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017687.mm.1 TC1700002811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017687.mm.1 NM_008202.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017687.mm.1 NM_008202.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024782.mm.1 TC1700002811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700024782.mm.1 NM_001077709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024782.mm.1 NM_001077709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024782.mm.1 NM_008202.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024782.mm.1 NM_008202.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017692.mm.1 TC1700002811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017692.mm.1 NM_001077709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017692.mm.1 NM_001077709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017692.mm.1 NM_008202.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017692.mm.1 NM_008202.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 462547 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0