Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00434316_m1
          See other FABP6 GE Assays ›
          Gene Symbol:
          Fabp6
          Gene Name:
          fatty acid binding protein 6, ileal (gastrotropin)
          Gene Aliases:
          GT, I-15P, I-BABP, ILBP, ILBP3, Illbp
          Chromosome Location:
          Chr.11: 43596040 - 43601562 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00434316_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fabp6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008375.2 NP_032401.1 3 - 4 383 63
          GenBank mRNA BC119289.1 - 3 - 4 353 8734038 63
          BC120767.1 - 3 - 4 353 40129690 63
          CJ043022.1 - 3 - 4 383 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002570.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fabp6

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          bile acid metabolic process
          transporter activity
          fatty acid binding
          lipid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033795 NM_008375.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033795 BC119289.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033795 BC120767.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554697 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150524 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033795 NM_008375.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033795 BC119289.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033795 BC120767.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27751411 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07322025 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150524 - -
          Whole Mouse Genome G4122A A_51_P309066 NM_008375.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P309066 BC119289.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P309066 BC120767.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104439 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150524 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08999198 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09415624 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09415629 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09415630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27751433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09205923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09170062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09181783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09403879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09246912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09405359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09041257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09159406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09544580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09581714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09744688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09403884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09336356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09577871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09614258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09226324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09287315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09061886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09675290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09336354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27751407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08999198 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10385340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385338 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385342 - -
          Affymetrix HT MG-430 PM Array Plate 1450682_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10385337 - -
          Mouse Genome 430 2.0 Array 1450682_at - -
          Mouse Genome 430 2.0 Array 1450682_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16204_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16204_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100023756.mm.1 TC1100002570.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100023756.mm.1 NM_008375.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023757.mm.1 TC1100002570.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100023757.mm.1 NM_008375.2 Refseq
          Mouse Expression Set 430 1450682_PM_at - -
          Mouse Expression Set 430 1450682_at - -
          Murine Genome U74v2 Set 99977_PM_at - -
          Murine Genome U74v2 Set 160886_PM_i_at - -
          Murine Genome U74v2 Set 99977_at - -
          Murine Genome U74v2 Set 160886_i_at - -
          Mouse Genome 430A 2.0 Array 1450682_PM_at - -
          Mouse Genome 430A 2.0 Array 1450682_at - -
          Affymetrix HT Mouse Genome 430A Array 1450682_at - -
          Affymetrix HT Mouse Genome 430A Array 1450682_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 508343 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7000278 - -
          Illumina mouse-6 v1.0 expression beadchip 7000278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227520 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227520 - -
          mouse-6 v1.1 (Illumina) scl016204.3_74-S - -
          Illumina Sentrix mouse-6.v1 bead array 7000278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl016204.3_74-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000278 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2826304 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2826304 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0