Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00434678_g1
          See other KLRA2 GE Assays ›
          Gene Symbol:
          Klra2
          Gene Name:
          killer cell lectin-like receptor, subfamily A, member 2
          Gene Aliases:
          Klra30, Ly49b
          Chromosome Location:
          Chr.6: 131198976 - 131247723 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00434678_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klra2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170851.1 NP_001164322.1 8 - 9 1093 139
          NM_008462.5 NP_032488.4 7 - 8 994 139
          GenBank mRNA AK017140.1 - 7 - 8 985 139
          BC064711.1 - 8 - 9 994 30295755 139
          U10304.1 - 7 - 8 922 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003264.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klra2

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004562 BC064711.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004562 NM_008462.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004562 NM_001170851.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 80617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267827 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 280283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184388 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 405437 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 522533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 280939 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186743 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 205794 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 18418 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168367 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148917 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138133 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004562 BC064711.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004562 NM_008462.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004562 NM_001170851.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P147466 BC064711.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P147466 EF025059.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P147466 NM_008462.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P147466 AF253058.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P147466 AF253059.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P269703 U10304.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P269703 AY860971.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P269703 NM_008462.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P269703 AF253057.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P269703 AF253058.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24850011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24849999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24850005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24850010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24850017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24850004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24850014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24850006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24850016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24850008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24850002 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138133 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148917 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06070879 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06070896 - -
          Whole Mouse Genome G4122A A_51_P269703 U10304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269703 AY860971.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269703 NM_008462.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269703 AF253057.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269703 AF253058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147466 BC064711.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147466 EF025059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147466 NM_008462.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147466 AF253058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147466 AF253059.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139745 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102104 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148917 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138133 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05870651 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05870652 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05870653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24850008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24850004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24850044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24850016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24850010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24850002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24850012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24850047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24850017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24850014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24850011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24850006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24849950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24850017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24849984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24849987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24850012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24850011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24849993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24849974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24849978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24849982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05209999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24850002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24849997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24849976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24849981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24849985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24849999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24850010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24850016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24850008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24849990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05551390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24849979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24850004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24850006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05870652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05209980 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548553 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548555 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548554 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548562 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548558 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548556 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548561 - -
          Affymetrix HT MG-430 PM Array Plate 1421304_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10548552 - -
          Mouse Genome 430 2.0 Array 1421304_at - -
          Mouse Genome 430 2.0 Array 1421304_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16633_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16633_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600025675.mm.1 TC0600003264.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025675.mm.1 NM_001170851.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025675.mm.1 NM_001170851.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025675.mm.1 NM_008462.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025675.mm.1 NM_008462.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025674.mm.1 TC0600003264.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025674.mm.1 NM_001170851.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025674.mm.1 NM_001170851.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025674.mm.1 NM_008462.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025674.mm.1 NM_008462.5 Refseq
          Mouse Expression Set 430 1421304_PM_at - -
          Mouse Expression Set 430 1421304_at - -
          Murine Genome U74v2 Set 98801_PM_at - -
          Murine Genome U74v2 Set 98801_at - -
          Mouse Genome 430A 2.0 Array 1421304_at - -
          Mouse Genome 430A 2.0 Array 1421304_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421304_at - -
          Affymetrix HT Mouse Genome 430A Array 1421304_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 819065 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 380102 - -
          Illumina mouse-6 v1.0 expression beadchip 380102 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000380102 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251941 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251941 - -
          mouse-6 v1.1 (Illumina) scl28315.5.1_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 380102 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28315.5.1_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 380102 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1251941 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2814974 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251941 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2814974 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0