Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00436844_m1
          See other STXBP1 GE Assays ›
          Gene Symbol:
          Stxbp1
          Gene Name:
          syntaxin binding protein 1
          Gene Aliases:
          AI317162, AI326233, MMS10-G, Ms10g, Munc-18a, Munc18-1, N-sec1, Rb-sec1, Sxtbp1, Unc18-1, Unc18h, nsec1
          Chromosome Location:
          Chr.2: 32787607 - 32847237 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00436844_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stxbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113569.1 NP_001107041.1 13 - 14 1259 128
          NM_009295.2 NP_033321.2 13 - 14 1254 128
          GenBank mRNA AF326545.1 - 13 - 14 1113 128
          AF326563.1 - 13 - 14 1113 128
          AK004780.1 - 8 - 9 978 128
          BC031728.1 - 13 - 14 1186 4480978 128
          D45903.1 - 13 - 14 1148 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003313.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stxbp1

          Gene Ontology Categories:

          Function(s) Process(es)

          platelet degranulation
          transport
          exocytosis
          vesicle docking involved in exocytosis
          neurotransmitter secretion
          neuromuscular synaptic transmission
          axon target recognition
          regulation of synaptic vesicle priming
          protein transport
          synaptic vesicle maturation
          vesicle-mediated transport
          synaptic vesicle fusion to presynaptic active zone membrane
          negative regulation of synaptic transmission, GABAergic
          negative regulation of neuron apoptotic process
          positive regulation of exocytosis
          positive regulation of calcium ion-dependent exocytosis
          protein stabilization
          long-term synaptic depression
          platelet aggregation
          protein localization to plasma membrane
          SNARE binding
          RNA binding
          protein binding
          syntaxin-1 binding
          protein kinase binding
          protein domain specific binding
          syntaxin binding
          identical protein binding
          protein N-terminus binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200028271.mm.1 TC0200003313.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028271.mm.1 NM_001113569.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028271.mm.1 NM_001113569.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028271.mm.1 NM_009295.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028271.mm.1 NM_009295.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
          git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline