Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00436930_g1
          See other TERT GE Assays ›
          Gene Symbol:
          Tert
          Gene Name:
          telomerase reverse transcriptase
          Gene Aliases:
          EST2, TCS1, TP2, TR, TRT
          Chromosome Location:
          Chr.13: 73626797 - 73649843 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00436930_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tert
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009354.1 NP_033380.1 1 - 2 249 54
          GenBank mRNA AF051911.1 - 1 - 2 249 54
          AF073311.1 - 1 - 2 220 54
          AK133783.1 - 1 - 2 340 54
          BC082327.1 - 1 - 2 260 54
          BC127068.1 - 1 - 2 239 40110173 54

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000885.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC127068.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tert

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, RNA-templated
          RNA-dependent DNA biosynthetic process
          telomere maintenance via telomerase
          mitochondrion organization
          negative regulation of gene expression
          DNA strand elongation
          positive regulation of Wnt signaling pathway
          production of siRNA involved in RNA interference
          RNA biosynthetic process
          positive regulation of hair cycle
          positive regulation of nitric-oxide synthase activity
          cellular response to hypoxia
          DNA biosynthetic process
          replicative senescence
          positive regulation of pri-miRNA transcription by RNA polymerase II
          negative regulation of production of siRNA involved in RNA interference
          positive regulation of stem cell proliferation
          negative regulation of cellular senescence
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          tRNA binding
          transcription coactivator binding
          DNA binding
          telomerase activity
          telomerase RNA reverse transcriptase activity
          RNA binding
          RNA-directed DNA polymerase activity
          RNA-directed 5'-3' RNA polymerase activity
          protein binding
          transferase activity
          nucleotidyltransferase activity
          telomeric DNA binding
          protein homodimerization activity
          metal ion binding
          telomerase RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300005691.mm.1 TC1300000885.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005691.mm.1 NM_009354.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005691.mm.1 NM_009354.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005690.mm.1 TC1300000885.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005690.mm.1 NM_009354.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005690.mm.1 NM_009354.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005692.mm.1 TC1300000885.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005692.mm.1 NM_009354.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005692.mm.1 NM_009354.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005693.mm.1 TC1300000885.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005693.mm.1 NM_009354.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005693.mm.1 NM_009354.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0