Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00436980_m1
          See other TGM2 GE Assays ›
          Gene Symbol:
          Tgm2
          Gene Name:
          transglutaminase 2, C polypeptide
          Gene Aliases:
          G[a]h, TG2, TGase2, tTG, tTGas
          Chromosome Location:
          Chr.2: 158116405 - 158146392 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00436980_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgm2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009373.3 NP_033399.1 3 - 4 530 67
          GenBank mRNA AF076928.1 - 2 - 3 445 67
          AK052912.1 - 3 - 4 505 67
          AK054108.1 - 3 - 4 505 67
          AK080224.1 - 3 - 4 530 67
          AK080593.1 - 3 - 4 505 67
          AK086001.1 - 3 - 4 505 67
          AK089481.1 - 3 - 4 533 67
          AK143712.1 - 3 - 4 566 67
          AK151776.1 - 3 - 4 531 67
          AK152152.1 - 3 - 4 505 67
          AK152627.1 - 3 - 4 531 67
          AK159255.1 - 3 - 4 505 67
          AK166302.1 - 3 - 4 505 67
          AK168990.1 - 3 - 4 506 67
          AK169356.1 - 3 - 4 533 67
          BC016492.1 - 3 - 4 471 3256943 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005030.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tgm2

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel remodeling
          G protein-coupled receptor signaling pathway
          phospholipase C-activating G protein-coupled receptor signaling pathway
          peptide cross-linking
          isopeptide cross-linking via N6-(L-isoglutamyl)-L-lysine
          negative regulation of endoplasmic reticulum calcium ion concentration
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          apoptotic cell clearance
          positive regulation of cell adhesion
          positive regulation of smooth muscle cell proliferation
          positive regulation of inflammatory response
          protein homooligomerization
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G protein-coupled signaling pathway
          positive regulation of mitochondrial calcium ion concentration
          branching involved in salivary gland morphogenesis
          salivary gland cavitation
          protein-glutamine gamma-glutamyltransferase activity
          calcium ion binding
          GTP binding
          transaminase activity
          transferase activity
          acyltransferase activity
          protein domain specific binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200043060.mm.1 TC0200005030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200043060.mm.1 NM_009373.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043060.mm.1 NM_009373.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043062.mm.1 TC0200005030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200043062.mm.1 NM_009373.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043062.mm.1 NM_009373.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043061.mm.1 TC0200005030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200043061.mm.1 NM_009373.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043061.mm.1 NM_009373.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 804123 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0