Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700026407.mm.1 |
TC0700002970.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026407.mm.1 |
NM_001285517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026407.mm.1 |
NM_001285518.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026407.mm.1 |
NM_001285519.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026407.mm.1 |
NM_009473.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026411.mm.1 |
TC0700002970.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026411.mm.1 |
NM_001285517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026411.mm.1 |
NM_001285518.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026411.mm.1 |
NM_001285519.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026411.mm.1 |
NM_009473.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026415.mm.1 |
TC0700002970.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026415.mm.1 |
NM_001285517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026415.mm.1 |
NM_001285518.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026415.mm.1 |
NM_001285519.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026415.mm.1 |
NM_009473.3 |
Refseq |