Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00438045_m1
          See other CASP3 GE Assays ›
          Gene Symbol:
          Casp3
          Gene Name:
          caspase 3
          Gene Aliases:
          A830040C14Rik, AC-3, Apopain, CASP-3, CC3, CPP-32, CPP32, Caspase-3, Lice, SCA-1, Yama, mldy
          Chromosome Location:
          Chr.8: 46617291 - 46639698 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00438045_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Casp3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009810.3 NP_033940.1 1 - 2 200 74
          GenBank mRNA U49929.1 - 1 - 2 42 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000541.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_009810.3,U49929.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protease Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Casp3

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          B cell homeostasis
          proteolysis
          apoptotic process
          cellular response to DNA damage stimulus
          heart development
          sensory perception of sound
          learning or memory
          response to UV
          response to wounding
          response to glucose
          protein processing
          neuron differentiation
          keratinocyte differentiation
          erythrocyte differentiation
          negative regulation of B cell proliferation
          glial cell apoptotic process
          intracellular signal transduction
          response to hydrogen peroxide
          T cell homeostasis
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          positive regulation of neuron apoptotic process
          cell fate commitment
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of cell cycle
          negative regulation of activated T cell proliferation
          neurotrophin TRK receptor signaling pathway
          neuron apoptotic process
          cellular response to organic substance
          execution phase of apoptosis
          protease binding
          aspartic-type endopeptidase activity
          cysteine-type endopeptidase activity
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          death receptor binding
          protein binding
          peptidase activity
          cysteine-type peptidase activity
          phospholipase A2 activator activity
          hydrolase activity
          protein complex binding
          cysteine-type endopeptidase activity involved in apoptotic process
          cysteine-type endopeptidase activity involved in execution phase of apoptosis

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800003835.mm.1 NM_001284409.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800003835.mm.1 NM_009810.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800003835.mm.1 TC0800000541.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 456269 - -
          ABI Mouse Genome Survey Microarray 561901 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0