Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00438289_g1
          See other COX4I1 GE Assays ›
          Gene Symbol:
          Cox4i1
          Gene Name:
          cytochrome c oxidase subunit IV isoform 1
          Gene Aliases:
          AL024441, COX, COX IV-1, COXIV, Cox4, Cox4a, IV-1
          Chromosome Location:
          Chr.8: 120668225 - 120674209 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00438289_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cox4i1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001293559.1 NP_001280488.1 3 - 4 299 61
          NM_009941.3 NP_034071.2 3 - 4 374 61
          GenBank mRNA AK002943.1 - 631 61
          AK012583.1 - 3 - 4 311 61
          AK019276.1 - 3 - 4 276 61
          AK150447.1 - 3 - 4 304 61
          AV469576.1 - 3 - 4 299 61
          BC132269.1 - 3 - 4 295 40130546 61
          BC132275.1 - 3 - 4 295 40130552 61
          M37829.1 - 3 - 4 297 61
          X54691.1 - 3 - 4 299 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001447.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cox4i1

          Gene Ontology Categories:

          Process(es)

          cytochrome-c oxidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26221739 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26221737 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26221738 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26221741 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165435 M37829.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165435 X54691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165435 AK002943.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165435 AK012583.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165435 NM_009941.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165435 M37829.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165435 X54691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165435 AK002943.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165435 AK012583.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165435 NM_009941.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288681 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 93299 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125985 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165435 M37829.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165435 X54691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165435 AK002943.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165435 AK012583.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165435 NM_009941.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165435 M37829.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165435 X54691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165435 AK002943.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165435 AK012583.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165435 NM_009941.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165435 M37829.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165435 X54691.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165435 AK002943.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165435 AK012583.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165435 NM_009941.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165435 M37829.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165435 X54691.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165435 AK002943.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165435 AK012583.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165435 NM_009941.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26221759 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125985 - -
          Whole Mouse Genome G4122A A_51_P165435 M37829.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165435 X54691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165435 AK002943.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165435 AK012583.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165435 NM_009941.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165435 M37829.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165435 X54691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165435 AK002943.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165435 AK012583.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165435 NM_009941.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111194 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125985 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07359886 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26221739 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26221741 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26221737 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26221738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26221820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07215708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07368896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07368905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06854807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07215706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07368902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07368908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06848919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26221764 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10576032 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576030 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576033 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576031 - -
          Affymetrix HT MG-430 PM Array Plate 1448322_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10576029 - -
          Mouse Genome 430 2.0 Array 1448322_a_at - -
          Mouse Genome 430 2.0 Array 1448322_PM_a_at - -
          Mouse Genome 430 2.0 Array 1434952_at - -
          Mouse Genome 430 2.0 Array 1434952_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12857_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12857_at - -
          Affymetrix Mouse Clariom S Array PSR0800011841.mm.1 TC0800001447.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800011841.mm.1 NM_009941.3 Refseq
          Mouse Expression Set 430 1448322_a_at - -
          Mouse Expression Set 430 1448322_PM_a_at - -
          Murine Genome U74v2 Set 93788_at - -
          Murine Genome U74v2 Set 102124_f_at - -
          Murine Genome U74v2 Set 93788_PM_at - -
          Murine Genome U74v2 Set 102124_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1448322_a_at - -
          Mouse Genome 430A 2.0 Array 1448322_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448322_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448322_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 910978 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5340427 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005340427 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249867 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249867 - -
          mouse-6 v1.1 (Illumina) scl33243.5.1_27-S - -
          Illumina Sentrix mouse-6.v1 bead array 5340427 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33243.5.1_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5340427 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2600274 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2600274 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl33243.5.1_27-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0