Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00438305_m1
          See other CRHR2 GE Assays ›
          Gene Symbol:
          Crhr2
          Gene Name:
          corticotropin releasing hormone receptor 2
          Gene Aliases:
          CRF-R2, CRFR2alpha, CRFR2beta, Crfr2
          Chromosome Location:
          Chr.6: 55090048 - 55133016 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00438305_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crhr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001288618.1 NP_001275547.1 3 - 4 468 65
          NM_001288619.1 NP_001275548.1 2 - 3 375 65
          NM_001288620.1 NP_001275549.1 2 - 3 375 65
          NM_009953.4 NP_034083.2 3 - 4 468 65
          GenBank mRNA AK085854.1 - 3 - 4 469 65
          AK142261.1 - 3 - 4 418 65
          AY445512.1 - 2 - 3 375 65
          AY753668.1 - 2 - 3 447 65
          BC137592.1 - 3 - 4 351 8860611 65
          CJ065172.1 - 3 - 4 418 65
          FJ548565.1 - 3 - 4 293 65
          U17858.1 - 3 - 4 293 65
          U19939.1 - 3 - 4 424 65
          U21729.1 - 3 - 4 419 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002325.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001288620.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crhr2

          Gene Ontology Categories:

          Function(s) Process(es)

          actin filament organization
          signal transduction
          cell surface receptor signaling pathway
          G protein-coupled receptor signaling pathway
          protein kinase C-activating G protein-coupled receptor signaling pathway
          hormone-mediated signaling pathway
          positive regulation of heart rate
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of norepinephrine secretion
          positive regulation of serotonin secretion
          negative regulation of angiogenesis
          sensory perception of pain
          epithelial cell differentiation
          positive regulation of interleukin-6 production
          negative regulation of epinephrine secretion
          positive regulation of stress-activated MAPK cascade
          negative regulation of luteinizing hormone secretion
          gastric motility
          catecholamine biosynthetic process
          positive regulation of cAMP-mediated signaling
          negative regulation of cAMP-mediated signaling
          positive regulation of blood pressure
          negative regulation of follicle-stimulating hormone secretion
          skeletal muscle tissue growth
          positive regulation of NF-kappaB transcription factor activity
          long-term synaptic potentiation
          negative regulation of insulin secretion involved in cellular response to glucose stimulus
          regulation of ERK1 and ERK2 cascade
          cellular response to corticotropin-releasing hormone stimulus
          negative regulation of calcium ion import
          negative regulation of feeding behavior
          negative regulation of defecation
          transmembrane signaling receptor activity
          G protein-coupled receptor activity
          hormone activity
          protein binding
          corticotrophin-releasing factor receptor activity
          peptide hormone binding
          corticotropin-releasing hormone receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600018178.mm.1 TC0600002325.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018178.mm.1 NM_001288618.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018178.mm.1 NM_001288619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018178.mm.1 NM_001288620.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018178.mm.1 NM_009953.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018181.mm.1 TC0600002325.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018181.mm.1 NM_001288619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018181.mm.1 NM_001288620.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018182.mm.1 TC0600002325.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018182.mm.1 NM_001288619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018182.mm.1 NM_001288620.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0