Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00438426_g1
          See other DLX2 GE Assays ›
          Gene Symbol:
          Dlx2
          Gene Name:
          distal-less homeobox 2
          Gene Aliases:
          AW121999, Dlx-2, Tes-1
          Chromosome Location:
          Chr.2: 71543408 - 71546754 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00438426_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dlx2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010054.2 NP_034184.1 1 - 2 770 67
          GenBank mRNA AK144652.1 - 1 - 2 771 67
          BC094317.1 - 1 - 2 470 5718422 67
          M80540.1 - 1 - 2 708 67
          M83184.1 - 1 - 2 333 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003728.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dlx2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          multicellular organism development
          proximal/distal pattern formation
          subpallium development
          hippocampus development
          olfactory bulb development
          forebrain neuron differentiation
          cerebral cortex GABAergic interneuron differentiation
          cerebral cortex GABAergic interneuron fate commitment
          cell differentiation
          odontogenesis of dentin-containing tooth
          negative regulation of Notch signaling pathway
          positive regulation of transcription by RNA polymerase II
          embryonic cranial skeleton morphogenesis
          embryonic skeletal system development
          negative regulation of oligodendrocyte differentiation
          branching morphogenesis of a nerve
          cartilage development
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          DNA-binding transcription activator activity, RNA polymerase II-specific
          DNA binding
          chromatin binding
          single-stranded RNA binding
          protein binding
          sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200031913.mm.1 TC0200003728.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200031913.mm.1 NM_010054.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031913.mm.1 NM_010054.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031914.mm.1 TC0200003728.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200031914.mm.1 NM_010054.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031914.mm.1 NM_010054.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031911.mm.1 TC0200003728.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200031911.mm.1 NM_010054.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031911.mm.1 NM_010054.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0