Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00438920_g1
          See other FGF8 GE Assays ›
          Gene Symbol:
          Fgf8
          Gene Name:
          fibroblast growth factor 8
          Gene Aliases:
          Aigf, Fgf-8
          Chromosome Location:
          Chr.19: 45736798 - 45742935 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00438920_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgf8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166361.1 NP_001159833.1 1 - 2 233 94
          NM_001166362.1 NP_001159834.1 1 - 2 233 94
          NM_001166363.1 NP_001159835.1 1 - 2 233 94
          NM_010205.2 NP_034335.1 1 - 2 233 94
          GenBank mRNA AK131980.1 - 1 - 2 209 94
          BC048734.1 - 1 - 2 223 6513131 94
          BU512456.1 - 1 - 2 268 94
          D12482.1 - 1 - 2 206 94
          D12483.1 - 1 - 2 232 94
          U18673.1 - 1 - 2 117 94
          Z48746.1 - 1 - 2 93 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001586.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK131980.1, BC048734.1, D12482.1, D12483.1, NM_001166361.1, NM_001166362.1, NM_001166363.1, NM_010205.2, U18673.1, Z48746.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fgf8

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          branching involved in blood vessel morphogenesis
          branching involved in ureteric bud morphogenesis
          organ induction
          kidney development
          neural plate morphogenesis
          heart looping
          blood vessel remodeling
          heart morphogenesis
          outflow tract septum morphogenesis
          outflow tract morphogenesis
          epithelial to mesenchymal transition involved in endocardial cushion formation
          apoptotic process
          response to oxidative stress
          multicellular organism development
          determination of left/right symmetry
          heart development
          motor neuron axon guidance
          mesodermal cell migration
          positive regulation of cell population proliferation
          gonad development
          fibroblast growth factor receptor signaling pathway
          embryo development ending in birth or egg hatching
          positive regulation of gene expression
          telencephalon development
          pallium development
          subpallium development
          forebrain dorsal/ventral pattern formation
          cell proliferation in forebrain
          forebrain neuron development
          central nervous system neuron development
          signal transduction involved in regulation of gene expression
          cell differentiation
          lung development
          BMP signaling pathway
          male genitalia development
          thyroid gland development
          otic vesicle formation
          midbrain-hindbrain boundary development
          dorsal/ventral axon guidance
          embryonic heart tube development
          limb morphogenesis
          embryonic hindlimb morphogenesis
          aorta morphogenesis
          inner ear morphogenesis
          regulation of odontogenesis of dentin-containing tooth
          negative regulation of apoptotic process
          negative regulation of neuron apoptotic process
          cell fate commitment
          positive regulation of mitotic nuclear division
          positive regulation of organ growth
          generation of neurons
          forebrain morphogenesis
          positive chemotaxis
          positive regulation of cell division
          negative regulation of cardiac muscle tissue development
          pharyngeal system development
          canonical Wnt signaling pathway
          corticotropin hormone secreting cell differentiation
          thyroid-stimulating hormone-secreting cell differentiation
          bone development
          lung morphogenesis
          branching involved in salivary gland morphogenesis
          neuroepithelial cell differentiation
          dopaminergic neuron differentiation
          cell migration involved in mesendoderm migration
          signaling receptor binding
          fibroblast growth factor receptor binding
          type 1 fibroblast growth factor receptor binding
          type 2 fibroblast growth factor receptor binding
          growth factor activity
          chemoattractant activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900014048.mm.1 TC1900001586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900014048.mm.1 NM_001166361.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014048.mm.1 NM_001166361.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014048.mm.1 NM_001166362.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014048.mm.1 NM_001166362.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014048.mm.1 NM_001166363.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014048.mm.1 NM_001166363.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014048.mm.1 NM_010205.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014048.mm.1 NM_010205.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014046.mm.1 TC1900001586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900014046.mm.1 NM_001166361.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014046.mm.1 NM_001166361.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014046.mm.1 NM_001166362.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014046.mm.1 NM_001166362.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014046.mm.1 NM_001166363.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014046.mm.1 NM_001166363.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014046.mm.1 NM_010205.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014046.mm.1 NM_010205.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014045.mm.1 TC1900001586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900014045.mm.1 NM_010205.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014045.mm.1 NM_010205.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014047.mm.1 TC1900001586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900014047.mm.1 NM_001166361.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014047.mm.1 NM_001166361.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014047.mm.1 NM_001166362.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014047.mm.1 NM_001166362.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014047.mm.1 NM_001166363.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014047.mm.1 NM_001166363.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014047.mm.1 NM_010205.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014047.mm.1 NM_010205.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0