Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00439635_m1
          See other IL4RA GE Assays ›
          Gene Symbol:
          Il4ra
          Gene Name:
          interleukin 4 receptor, alpha
          Gene Aliases:
          CD124, Il4r
          Chromosome Location:
          Chr.7: 125552282 - 125579474 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00439635_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il4ra
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008700.3 NP_001008700.1 2 - 3 227 121
          GenBank mRNA AK034466.1 - 2 - 3 271 121
          AK042342.1 - 2 - 3 227 121
          AK088086.1 - 2 - 3 239 121
          AK137827.1 - 2 - 3 239 121
          AK149603.1 - 2 - 3 240 121
          AK149717.1 - 2 - 3 240 121
          AK150584.1 - 2 - 3 239 121
          AK151998.1 - 2 - 3 239 121
          AK153186.1 - 2 - 3 239 121
          AK159375.1 - 2 - 3 238 121
          AK160016.1 - 2 - 3 231 121
          BC112911.1 - 2 - 3 227 30138743 121
          BC128301.1 - 1 - 2 98 121
          BC128302.1 - 1 - 2 98 121
          M27959.1 - 2 - 3 222 121
          M27960.1 - 2 - 3 222 121
          M29854.1 - 2 - 3 206 121

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK034466.1, AK042342.1, AK088086.1, AK149603.1, AK149717.1, AK150584.1, AK151998.1, AK153186.1, AK159375.1, AK160016.1, BC112911.1, BC128301.1, BC128302.1, M27959.1, M27960.1, M29854.1, NM_001008700.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il4ra

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          production of molecular mediator involved in inflammatory response
          positive regulation of immunoglobulin production
          immunoglobulin mediated immune response
          positive regulation of chemokine production
          regulation of cell population proliferation
          defense response to protozoan
          positive regulation of macrophage activation
          positive regulation of mast cell degranulation
          negative regulation of T-helper 1 cell differentiation
          positive regulation of T-helper 2 cell differentiation
          positive regulation of myoblast fusion
          cytokine receptor activity
          protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0