Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00440843_mH
          See other PRKACB GE Assays ›
          Gene Symbol:
          Prkacb
          Gene Name:
          protein kinase, cAMP dependent, catalytic, beta
          Gene Aliases:
          Pkacb
          Chromosome Location:
          Chr.3: 146729579 - 146812946 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00440843_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prkacb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164198.1 NP_001157670.1 8 - 9 1021 82
          NM_001164199.1 NP_001157671.1 8 - 9 894 82
          NM_001164200.1 NP_001157672.1 8 - 9 986 82
          NM_011100.4 NP_035230.1 8 - 9 1025 82
          GenBank mRNA AK035560.1 - 1 - 2 109 82
          AK048319.1 - 8 - 9 1017 82
          AK139419.1 - 8 - 9 766 82
          AK148728.1 - 8 - 9 835 82
          AK150122.1 - 8 - 9 987 82
          AK150134.1 - 8 - 9 987 82
          AK163585.1 - 8 - 9 799 82
          BC054533.1 - 8 - 9 1009 5704796 82
          J02626.1 - 8 - 9 954 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300003106.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prkacb

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          protein phosphorylation
          adenylate cyclase-modulating G protein-coupled receptor signaling pathway
          phosphorylation
          negative regulation of meiotic cell cycle
          regulation of protein processing
          negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
          nucleotide binding
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          cAMP-dependent protein kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          ubiquitin protein ligase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300024445.mm.1 TC0300003106.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024445.mm.1 NM_001164198.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024445.mm.1 NM_001164198.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024445.mm.1 NM_001164199.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024445.mm.1 NM_001164199.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024445.mm.1 NM_001164200.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024445.mm.1 NM_001164200.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024445.mm.1 NM_011100.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024445.mm.1 NM_011100.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline