Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00441459_m1
          See other SLC1A2 GE Assays ›
          Gene Symbol:
          Slc1a2
          Gene Name:
          solute carrier family 1 (glial high affinity glutamate transporter), member 2
          Gene Aliases:
          1700091C19Rik, 2900019G14Rik, AI159670, Eaat2, GLT-1, GLT1, MGLT1
          Chromosome Location:
          Chr.2: 102658594 - 102790784 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00441459_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc1a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077514.3 NP_001070982.1 6 - 7 1451 101
          NM_001077515.2 NP_001070983.1 6 - 7 878 101
          NM_011393.2 NP_035523.1 6 - 7 878 101
          GenBank mRNA AB007810.1 - 6 - 7 1508 101
          AB007811.1 - 6 - 7 867 101
          AB007812.1 - 6 - 7 878 101
          AK134609.1 - 6 - 7 1452 101
          AK140095.1 - 6 - 7 1446 101
          BC138255.1 - 6 - 7 878 8861274 101
          BC138256.1 - 6 - 7 878 8861275 101
          D43796.1 - 6 - 7 1031 101
          KC825358.1 - 5 - 6 840 101
          U11763.1 - 6 - 7 1036 101
          U24699.1 - 5 - 6 849 101
          U75372.1 - 6 - 7 1464 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001524.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc1a2

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          dicarboxylic acid transport
          nervous system development
          visual behavior
          response to xenobiotic stimulus
          response to light stimulus
          response to wounding
          multicellular organism aging
          L-glutamate transmembrane transport
          telencephalon development
          adult behavior
          cellular response to extracellular stimulus
          multicellular organism growth
          sodium ion transmembrane transport
          response to amino acid
          positive regulation of glucose import
          L-glutamate import
          D-aspartate import across plasma membrane
          L-glutamate transmembrane transporter activity
          high-affinity glutamate transmembrane transporter activity
          symporter activity
          glutamate:sodium symporter activity
          sodium:dicarboxylate symporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200011452.mm.1 TC0200001524.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200011452.mm.1 NM_001077514.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011452.mm.1 NM_001077514.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011452.mm.1 NM_001077515.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011452.mm.1 NM_001077515.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011452.mm.1 NM_011393.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011452.mm.1 NM_011393.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011453.mm.1 TC0200001524.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200011453.mm.1 NM_001077514.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011453.mm.1 NM_001077514.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011453.mm.1 NM_001077515.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011453.mm.1 NM_001077515.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011453.mm.1 NM_011393.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011453.mm.1 NM_011393.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-9267x:80/100.66.134.224:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0