Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00443048_g1
          See other SLC11A1 GE Assays ›
          Gene Symbol:
          Slc11a1
          Gene Name:
          solute carrier family 11 (proton-coupled divalent metal ion transporters), member 1
          Gene Aliases:
          Bcg, Ity, Lsh, Nramp, Nramp1, ity
          Chromosome Location:
          Chr.1: 74375014 - 74386057 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00443048_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc11a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013612.2 NP_038640.2 7 - 8 739 99
          GenBank mRNA AK090303.1 - 7 - 8 740 99
          AK144237.1 - 7 - 8 722 99
          AK171343.1 - 6 - 7 975 99
          AK171393.1 - 7 - 8 720 99
          BC109137.1 - 7 - 8 653 40039390 99
          BC109138.1 - 7 - 8 653 40039397 99
          L13732.1 - 7 - 8 717 99
          X75355.1 - 7 - 8 698 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000632.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc11a1

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          negative regulation of cytokine production
          positive regulation of cytokine production
          T cell proliferation involved in immune response
          T cell cytokine production
          positive regulation of dendritic cell antigen processing and presentation
          positive regulation of T-helper 1 type immune response
          transport
          ion transport
          iron ion transport
          manganese ion transport
          cellular cadmium ion homeostasis
          cellular iron ion homeostasis
          phagocytosis
          inflammatory response
          vacuolar acidification
          response to bacterium
          positive regulation of gene expression
          nitrite transport
          activation of protein kinase activity
          response to lipopolysaccharide
          interleukin-2 production
          interleukin-3 production
          positive regulation of interferon-gamma production
          response to interferon-gamma
          wound healing
          macrophage activation
          defense response to bacterium
          defense response to protozoan
          MHC class II biosynthetic process
          respiratory burst
          positive regulation of transcription by RNA polymerase II
          antigen processing and presentation of peptide antigen
          mRNA stabilization
          positive regulation of phagocytosis
          defense response to Gram-negative bacterium
          iron ion homeostasis
          multicellular organismal iron ion homeostasis
          cadmium ion transmembrane transport
          metal ion export
          manganese ion transmembrane transport
          L-arginine transport
          proton transmembrane transport
          transporter activity
          manganese ion transmembrane transporter activity
          protein homodimerization activity
          transition metal ion transmembrane transporter activity
          metal ion:proton antiporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100005090.mm.1 TC0100000632.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100005090.mm.1 NM_013612.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100005090.mm.1 NM_013612.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0