Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00444533_m1
          See other CXCL13 GE Assays ›
          Gene Symbol:
          Cxcl13
          Gene Name:
          chemokine (C-X-C motif) ligand 13
          Gene Aliases:
          4631412M08Rik, ANGIE2, Angie, BCA-1, BLC, BLR1L, Scyb13
          Chromosome Location:
          Chr.5: 95956939 - 95961068 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00444533_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cxcl13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_018866.2 NP_061354.1 1 - 2 95 84
          GenBank mRNA AF030636.1 - 1 - 2 91 84
          AF044196.1 - 1 - 2 95 84
          AK156025.1 - 1 - 2 109 84
          BB612247.1 - 106 84
          BC012965.1 - 1 - 2 94 4217539 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000922.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Mouse Chemokine Panel Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cxcl13

          Gene Ontology Categories:

          Function(s) Process(es)

          lymphocyte chemotaxis across high endothelial venule
          positive regulation of leukocyte chemotaxis
          chemotaxis
          inflammatory response
          immune response
          cell surface receptor signaling pathway
          G protein-coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          positive regulation of T cell chemotaxis
          response to lipopolysaccharide
          positive regulation of integrin activation
          positive regulation of cell-cell adhesion mediated by integrin
          B cell chemotaxis
          endothelial cell chemotaxis to fibroblast growth factor
          B cell chemotaxis across high endothelial venule
          regulation of cell population proliferation
          defense response to bacterium
          lymph node development
          chemokine-mediated signaling pathway
          activation of GTPase activity
          positive regulation of signaling receptor activity
          negative regulation of endothelial cell chemotaxis to fibroblast growth factor
          cytokine activity
          chemokine activity
          heparin binding
          fibroblast growth factor binding
          CXCR5 chemokine receptor binding
          CCR10 chemokine receptor binding
          protein heterodimerization activity
          receptor ligand activity
          CXCR3 chemokine receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500007944.mm.1 TC0500000922.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007944.mm.1 NM_018866.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007944.mm.1 NM_018866.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007945.mm.1 TC0500000922.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007945.mm.1 NM_018866.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007945.mm.1 NM_018866.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0