Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00444842_g1
          See other HTRA2 GE Assays ›
          Gene Symbol:
          Htra2
          Gene Name:
          HtrA serine peptidase 2
          Gene Aliases:
          AI481710, Omi, Prss25, mnd2
          Chromosome Location:
          Chr.6: 83051266 - 83054571 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00444842_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Htra2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019752.3 NP_062726.3 1 - 2 649 83
          GenBank mRNA AF164513.1 - 1 - 2 528 83
          AF175324.1 - 1 - 2 545 83
          AK146338.1 - 1 - 2 540 83
          AK159188.1 - 1 - 2 469 83
          BC033276.1 - 1 - 2 509 83
          BC049880.1 - 1 - 2 649 5321861 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002555.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Htra2

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          apoptotic process
          mitochondrion organization
          adult walking behavior
          adult locomotory behavior
          intrinsic apoptotic signaling pathway in response to DNA damage
          positive regulation of cell death
          forebrain development
          cellular response to heat
          cellular response to interferon-beta
          regulation of multicellular organism growth
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular protein catabolic process
          neuron development
          negative regulation of cell death
          cellular response to retinoic acid
          cellular response to growth factor stimulus
          intrinsic apoptotic signaling pathway
          positive regulation of protein targeting to mitochondrion
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          serine-type endopeptidase activity
          protein binding
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600019798.mm.1 TC0600002555.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019798.mm.1 NM_019752.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019798.mm.1 NM_019752.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019799.mm.1 TC0600002555.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019799.mm.1 NM_019752.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019799.mm.1 NM_019752.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019794.mm.1 TC0600002555.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019794.mm.1 NM_019752.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019794.mm.1 NM_019752.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019795.mm.1 TC0600002555.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019795.mm.1 NM_019752.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019795.mm.1 NM_019752.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019800.mm.1 TC0600002555.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019800.mm.1 NM_019752.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019800.mm.1 NM_019752.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 448904 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0