Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00445223_g1
          See other KIT GE Assays ›
          Gene Symbol:
          Kit
          Gene Name:
          kit oncogene
          Gene Aliases:
          Bs, CD117, Fdc, Gsfsco1, Gsfsco5, Gsfsow3, SCO1, SCO5, SOW3, Ssm, Tr-kit, W, c-KIT
          Chromosome Location:
          Chr.5: 75574987 - 75656722 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00445223_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kit
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001122733.1 NP_001116205.1 19 - 20 2773 105
          NM_021099.3 NP_066922.2 19 - 20 2757 105
          GenBank mRNA AK046795.1 - 19 - 20 2765 105
          AK047010.1 - 19 - 20 2770 105
          AK145462.1 - 16 - 17 1998 105
          AY536430.1 - 19 - 20 2720 105
          AY536431.1 - 19 - 20 2720 105
          BC026713.1 - 19 - 20 2752 105
          BC052457.1 - 19 - 20 2732 6830637 105
          BC075716.1 - 19 - 20 2758 3673641 105
          EF472963.1 - 3 - 4 377 105
          X65997.1 - 3 - 4 792 105
          Y00864.1 - 19 - 20 2724 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000710.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kit

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          myeloid progenitor cell differentiation
          lymphoid progenitor cell differentiation
          immature B cell differentiation
          dendritic cell cytokine production
          mast cell chemotaxis
          myeloid leukocyte differentiation
          protein phosphorylation
          glycosphingolipid metabolic process
          chemotaxis
          inflammatory response
          transmembrane receptor protein tyrosine kinase signaling pathway
          germ cell development
          spermatogenesis
          spermatid development
          positive regulation of cell population proliferation
          germ cell migration
          regulation of cell shape
          visual learning
          response to radiation
          positive regulation of gene expression
          phosphorylation
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          lamellipodium assembly
          hemopoiesis
          cell differentiation
          T cell differentiation
          erythrocyte differentiation
          melanocyte differentiation
          positive regulation of cell migration
          positive regulation of pseudopodium assembly
          actin cytoskeleton reorganization
          mast cell cytokine production
          somatic stem cell population maintenance
          embryonic hemopoiesis
          ectopic germ cell programmed cell death
          intracellular signal transduction
          megakaryocyte development
          Fc receptor signaling pathway
          Kit signaling pathway
          erythropoietin-mediated signaling pathway
          positive regulation of tyrosine phosphorylation of STAT protein
          negative regulation of programmed cell death
          mast cell degranulation
          positive regulation of MAP kinase activity
          positive regulation of MAPK cascade
          pigmentation
          positive regulation of Notch signaling pathway
          positive regulation of receptor signaling pathway via JAK-STAT
          protein autophosphorylation
          developmental pigmentation
          regulation of developmental pigmentation
          somatic stem cell division
          positive regulation of long-term neuronal synaptic plasticity
          digestive tract development
          stem cell differentiation
          epithelial cell proliferation
          detection of mechanical stimulus involved in sensory perception of sound
          positive regulation of DNA-binding transcription factor activity
          cell chemotaxis
          mast cell differentiation
          cellular response to thyroid hormone stimulus
          melanocyte migration
          melanocyte adhesion
          nucleotide binding
          protease binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          stem cell factor receptor activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          cytokine binding
          protein homodimerization activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500006313.mm.1 TC0500000710.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500006313.mm.1 NM_001122733.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500006313.mm.1 NM_001122733.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500006313.mm.1 NM_021099.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500006313.mm.1 NM_021099.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0