Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00446294_m1
          See other NGFR GE Assays ›
          Gene Symbol:
          Ngfr
          Gene Name:
          nerve growth factor receptor (TNFR superfamily, member 16)
          Gene Aliases:
          LNGFR, Tnfrsf16, p75, p75NGFR, p75NTR
          Chromosome Location:
          Chr.11: 95568818 - 95587698 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00446294_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ngfr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033217.3 NP_150086.2 2 - 3 352 121
          GenBank mRNA AF105292.1 - 2 - 3 210 121
          AK037248.1 - 2 - 3 353 121
          BB630380.1 - 2 - 3 353 121
          BC038365.1 - 2 - 3 343 5367638 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003593.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ngfr

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          apoptotic process
          signal transduction
          multicellular organism development
          nervous system development
          axon guidance
          central nervous system development
          response to wounding
          regulation of gene expression
          positive regulation of cell death
          negative regulation of neuron projection development
          programmed cell death
          detection of temperature stimulus
          sensory perception of pain
          nerve development
          cell differentiation
          hair follicle morphogenesis
          positive regulation of myelination
          positive regulation of synaptic transmission, cholinergic
          circadian regulation of gene expression
          cellular response to oxidative stress
          positive regulation of Rho protein signal transduction
          negative regulation of fibroblast growth factor receptor signaling pathway
          positive regulation of odontogenesis of dentin-containing tooth
          glucose homeostasis
          positive regulation of apoptotic process
          positive regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          skin development
          positive regulation of fibroblast proliferation
          rhythmic process
          negative regulation of hair follicle development
          positive regulation of protein kinase B signaling
          negative regulation of mitochondrial depolarization
          positive regulation of synaptic transmission, glutamatergic
          negative regulation of cell death
          positive regulation of neuron death
          regulation of reactive oxygen species metabolic process
          positive regulation of apoptotic signaling pathway
          amyloid-beta binding
          death receptor activity
          neurotrophin TRKA receptor binding
          protein binding
          ubiquitin protein ligase binding
          neurotrophin binding
          nerve growth factor binding
          preprotein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100032952.mm.1 TC1100003593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100032952.mm.1 NM_033217.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032952.mm.1 NM_033217.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline