Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00446500_g1
          See other TIRAP GE Assays ›
          Gene Symbol:
          Tirap
          Gene Name:
          toll-interleukin 1 receptor (TIR) domain-containing adaptor protein
          Gene Aliases:
          AA407980, C130027E04Rik, Mal, Tlr4ap, Wyatt
          Chromosome Location:
          Chr.9: 35184391 - 35200291 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00446500_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tirap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177845.1 NP_001171316.1 3 - 4 344 67
          NM_001177846.1 NP_001171317.1 3 - 4 326 67
          NM_001177847.1 NP_001171318.1 3 - 4 272 67
          NM_054096.2 NP_473437.2 3 - 4 336 67
          GenBank mRNA AF378130.1 - 3 - 4 245 67
          AF410784.1 - 2 - 3 168 67
          AK036431.1 - 3 - 4 265 67
          AK080135.1 - 3 - 4 345 67
          AK151117.1 - 3 - 4 274 67
          AK168494.1 - 3 - 4 264 67
          BY740695.1 - 3 - 4 332 67
          BY743490.1 - 3 - 4 272 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002008.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AF378130.1, AF410784.1, AK036431.1, AK080135.1, AK151117.1, AK168494.1, NM_001177845.1, NM_001177846.1, NM_001177847.1, NM_054096.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tirap

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          inflammatory response
          signal transduction
          cell surface receptor signaling pathway
          I-kappaB kinase/NF-kappaB signaling
          response to bacterium
          myeloid cell differentiation
          positive regulation of B cell proliferation
          response to lipopolysaccharide
          regulation of interferon-beta production
          regulation of interleukin-15 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-15 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          positive regulation of toll-like receptor 2 signaling pathway
          positive regulation of toll-like receptor 3 signaling pathway
          positive regulation of toll-like receptor 4 signaling pathway
          toll-like receptor 5 signaling pathway
          TIRAP-dependent toll-like receptor 4 signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          innate immune response
          regulation of innate immune response
          positive regulation of innate immune response
          positive regulation of JNK cascade
          defense response to Gram-positive bacterium
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of ERK1 and ERK2 cascade
          interleukin-1-mediated signaling pathway
          3'-UTR-mediated mRNA stabilization
          cellular response to bacterial lipopeptide
          cellular response to lipoteichoic acid
          positive regulation of neutrophil chemotaxis
          positive regulation of protein homodimerization activity
          positive regulation of chemokine (C-X-C motif) ligand 1 production
          positive regulation of chemokine (C-X-C motif) ligand 2 production
          protein kinase C binding
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          Toll-like receptor 4 binding
          Toll-like receptor 2 binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900016676.mm.1 TC0900002008.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900016676.mm.1 NM_001177845.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016676.mm.1 NM_001177845.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016676.mm.1 NM_001177846.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016676.mm.1 NM_001177846.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016676.mm.1 NM_001177847.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016676.mm.1 NM_001177847.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016676.mm.1 NM_054096.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016676.mm.1 NM_054096.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0