Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00447493_g1
          See other TGFB1I1 GE Assays ›
          Gene Symbol:
          Tgfb1i1
          Gene Name:
          transforming growth factor beta 1 induced transcript 1
          Gene Aliases:
          ARA55, Hic5, TSC-5, hic-5
          Chromosome Location:
          Chr.7: 128246799 - 128255699 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00447493_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgfb1i1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289550.1 NP_001276479.1 3 - 4 363 71
          NM_001289551.1 NP_001276480.1 3 - 4 363 71
          NM_001289553.1 NP_001276482.1 2 - 3 247 71
          GenBank mRNA AK003170.1 - 3 - 4 198 71
          AK041233.1 - 3 - 4 191 71
          AK085042.1 - 2 - 3 89 71
          AK087114.1 - 3 - 4 198 71
          AK156423.1 - 3 - 4 198 71
          BC002049.1 - 3 - 4 197 3489459 71
          BC056362.1 - 2 - 3 1472 6816930 71
          DQ143891.1 - 2 - 3 248 71
          DQ143892.1 - 1 - 2 63 71
          DQ143895.1 - 1 - 2 452 71
          DQ143897.1 - 2 - 3 325 71
          DQ143898.1 - 2 - 3 325 71
          DQ143899.1 - 1 - 2 452 71
          L22482.1 - 2 - 3 324 71

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the DQ143892.1, DQ143895.1, DQ143897.1, DQ143898.1, DQ143899.1, NM_001289553.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          response to heat
          positive regulation of epithelial to mesenchymal transition
          Wnt signaling pathway
          morphogenesis of embryonic epithelium
          cell differentiation
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of transforming growth factor beta receptor signaling pathway
          ubiquitin-dependent SMAD protein catabolic process
          epithelial cell differentiation
          cell fate commitment
          negative regulation of fat cell differentiation
          positive regulation of transcription, DNA-templated
          transcription coactivator activity
          protein binding
          zinc ion binding
          metal ion binding
          Roundabout binding
          androgen receptor binding
          I-SMAD binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700016387.mm.1 TC0700001879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700016387.mm.1 NM_001289550.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016387.mm.1 NM_001289551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016387.mm.1 NM_001289553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016388.mm.1 TC0700001879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700016388.mm.1 NM_001289550.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016388.mm.1 NM_001289551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016388.mm.1 NM_001289553.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 493439 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0