Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00447497_m1
          See other TGFB1I1 GE Assays ›
          Gene Symbol:
          Tgfb1i1
          Gene Name:
          transforming growth factor beta 1 induced transcript 1
          Gene Aliases:
          ARA55, Hic5, TSC-5, hic-5
          Chromosome Location:
          Chr.7: 128246799 - 128255699 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00447497_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgfb1i1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289550.1 NP_001276479.1 7 - 8 899 90
          NM_001289552.1 NP_001276481.1 6 - 7 756 90
          NM_001289553.1 NP_001276482.1 6 - 7 783 90
          GenBank mRNA AK003170.1 - 7 - 8 733 90
          AK041233.1 - 7 - 8 728 90
          AK085042.1 - 6 - 7 625 90
          AK156423.1 - 7 - 8 734 90
          AK158409.1 - 6 - 7 608 90
          BC002049.1 - 7 - 8 3489459 90
          BC056362.1 - 6 - 7 2008 6816930 90
          DQ143891.1 - 6 - 7 784 90
          DQ143892.1 - 5 - 6 570 90
          DQ143893.1 - 5 - 6 577 90
          DQ143894.1 - 5 - 6 572 90
          DQ143895.1 - 5 - 6 988 90
          DQ143896.1 - 5 - 6 949 90
          DQ143897.1 - 5 - 6 1027 90
          DQ143898.1 - 5 - 6 976 90
          DQ143899.1 - 4 - 5 1103 90
          DQ143900.1 - 6 - 7 796 90
          L22482.1 - 6 - 7 860 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001879.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Mouse Androgen Panel 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          response to heat
          positive regulation of epithelial to mesenchymal transition
          Wnt signaling pathway
          morphogenesis of embryonic epithelium
          cell differentiation
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of transforming growth factor beta receptor signaling pathway
          ubiquitin-dependent SMAD protein catabolic process
          epithelial cell differentiation
          cell fate commitment
          negative regulation of fat cell differentiation
          positive regulation of transcription, DNA-templated
          transcription coactivator activity
          protein binding
          zinc ion binding
          metal ion binding
          Roundabout binding
          androgen receptor binding
          I-SMAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000122 AK041233.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000122 AK085042.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000127 DQ143899.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000127 DQ143900.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000127 L22482.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000127 DQ143896.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000127 DQ143897.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764806 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 88149 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167964 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000127 DQ143899.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000127 DQ143900.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000127 L22482.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000127 DQ143896.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000127 DQ143897.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P209383 DQ143899.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P209383 DQ143900.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P209383 L22482.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P209383 DQ143896.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P209383 DQ143897.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P366344 DQ143899.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P366344 DQ143900.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P366344 L22482.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P366344 DQ143896.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P366344 DQ143897.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25509716 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01632607 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167964 - -
          Whole Mouse Genome G4122A A_52_P949440 - -
          Whole Mouse Genome G4122A A_52_P209383 DQ143899.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P209383 DQ143900.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P209383 L22482.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P209383 DQ143896.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P209383 DQ143897.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P366344 DQ143899.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P366344 DQ143900.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P366344 L22482.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P366344 DQ143896.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P366344 DQ143897.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131915 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112667 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167964 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06523177 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06523188 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06523173 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06523176 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06523179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25509670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06523189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06271697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06271693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06655018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06523173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06523184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06560486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06602165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06523170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06346039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06274939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06271717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06523176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06274938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25509741 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10557966 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557969 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557964 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557963 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557967 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557962 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557971 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557972 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557961 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557965 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557970 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557968 - -
          Affymetrix HT MG-430 PM Array Plate 1418136_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10557960 - -
          Mouse Genome 430 2.0 Array 1418136_at - -
          Mouse Genome 430 2.0 Array 1418136_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21804_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21804_at - -
          Affymetrix Murine Genome 6500A Array Msa14180_at - -
          Affymetrix Mouse Clariom S Array PSR0700016396.mm.1 TC0700001879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700016396.mm.1 NM_001289550.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016396.mm.1 NM_001289551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016396.mm.1 NM_001289552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016396.mm.1 NM_001289553.1 Refseq
          Mouse Expression Set 430 1418136_at - -
          Mouse Expression Set 430 1418136_PM_at - -
          Murine Genome U74v2 Set 98019_at - -
          Murine Genome U74v2 Set 98019_PM_at - -
          Mouse Genome 430A 2.0 Array 1418136_PM_at - -
          Mouse Genome 430A 2.0 Array 1418136_at - -
          Affymetrix HT Mouse Genome 430A Array 1418136_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418136_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 493439 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2060288 - -
          Illumina mouse-6 v1.0 expression beadchip 6550450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002060288 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227722 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2693246 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2693246 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227722 - -
          mouse-6 v1.1 (Illumina) scl021804.11_211-S - -
          mouse-6 v1.1 (Illumina) scl0021804.1_159-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550450 - -
          Illumina Sentrix mouse-6.v1 bead array 2060288 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0021804.1_159-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl021804.11_211-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2060288 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550450 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2693246 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1227722 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227722 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2693246 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0021804.1_159-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl021804.11_211-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0