Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00447973_g1
          See other SEPT2 GE Assays ›
          Gene Symbol:
          Sept2
          Gene Name:
          septin 2
          Gene Aliases:
          AW208991, Nedd-5, Nedd5, mKIAA0158
          Chromosome Location:
          Chr.1: 93478967 - 93509733 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00447973_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sept2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159717.1 NP_001153189.1 6 - 7 572 134
          NM_001159718.1 NP_001153190.1 5 - 6 545 134
          NM_001159719.1 NP_001153191.1 6 - 7 620 134
          NM_010891.2 NP_035021.1 6 - 7 576 134
          GenBank mRNA AK028072.1 - 6 - 7 558 134
          AK146616.1 - 6 - 7 573 134
          AK151591.1 - 6 - 7 560 134
          AK171331.1 - 6 - 7 555 134
          BC138636.1 - 5 - 6 513 8861656 134
          BC138637.1 - 5 - 6 513 8861657 134
          D49382.2 - 6 - 7 531 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000936.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sept2

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          cytokinesis
          regulation of L-glutamate import across plasma membrane
          cell cycle
          smoothened signaling pathway
          neuron projection development
          regulation of protein localization
          regulation of catalytic activity
          protein polymerization
          cell division
          cilium assembly
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          enzyme regulator activity
          identical protein binding
          molecular adaptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100007960.mm.1 TC0100000936.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100007960.mm.1 NM_001159717.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007960.mm.1 NM_001159719.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007960.mm.1 NM_001159717.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007960.mm.1 NM_001159718.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007960.mm.1 NM_001159718.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007960.mm.1 NM_001159719.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007960.mm.1 NM_010891.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007960.mm.1 NM_010891.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007959.mm.1 TC0100000936.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100007959.mm.1 NM_001159717.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007959.mm.1 NM_001159717.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007959.mm.1 NM_001159718.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007959.mm.1 NM_001159718.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007959.mm.1 NM_001159719.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007959.mm.1 NM_001159719.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007959.mm.1 NM_010891.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007959.mm.1 NM_010891.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0