Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00447998_m1
          See other NRF1 GE Assays ›
          Gene Symbol:
          Nrf1
          Gene Name:
          nuclear respiratory factor 1
          Gene Aliases:
          C87038, D6Ertd415e
          Chromosome Location:
          Chr.6: 30047988 - 30153458 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00447998_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nrf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164226.1 NP_001157698.1 6 - 7 904 85
          NM_001164227.1 NP_001157699.1 6 - 7 904 85
          NM_001164228.1 NP_001157700.1 6 - 7 901 85
          NM_001164229.1 NP_001157701.1 7 - 8 978 85
          NM_001164230.1 NP_001157702.1 6 - 7 901 85
          NM_010938.4 NP_035068.3 6 - 7 901 85
          GenBank mRNA AF098077.1 - 6 - 7 836 85
          AK014494.1 - 6 - 7 865 85
          AK029034.1 - 6 - 7 882 85
          AK031103.1 - 1 - 2 105 85
          AK037697.1 - 6 - 7 862 85
          AK050861.1 - 6 - 7 818 85
          AK082580.1 - 6 - 7 902 85
          AK135673.1 - 7 - 8 981 85
          BC005410.1 - 5 - 6 722 3496661 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000257.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nrf1

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          mitochondrion organization
          positive regulation of transcription by RNA polymerase II
          response to electrical stimulus
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600001835.mm.1 TC0600000257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600001835.mm.1 NM_001164226.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001835.mm.1 NM_001164226.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001835.mm.1 NM_001164227.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001835.mm.1 NM_001164227.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001835.mm.1 NM_001164228.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001835.mm.1 NM_001164228.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001835.mm.1 NM_001164229.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001835.mm.1 NM_001164229.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001835.mm.1 NM_001164230.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001835.mm.1 NM_001164230.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001835.mm.1 NM_010938.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001835.mm.1 NM_010938.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0