Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00448086_m1
          See other SIGMAR1 GE Assays ›
          Gene Symbol:
          Sigmar1
          Gene Name:
          sigma non-opioid intracellular receptor 1
          Gene Aliases:
          Oprs1, sigma1R
          Chromosome Location:
          Chr.4: 41738493 - 41741359 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00448086_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sigmar1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286541.1 NP_001273470.1 2 - 3 353 72
          NM_001286551.1 NP_001273480.1 3 - 4 507 72
          NM_011014.3 NP_035144.1 3 - 4 554 72
          GenBank mRNA AF004927.1 - 3 - 4 508 72
          AF030198.1 - 3 - 4 468 72
          AK002718.1 - 3 - 4 487 72
          AK154300.1 - 3 - 4 526 72
          AK159886.1 - 3 - 4 488 72
          AY390768.1 - 2 - 3 246 72
          BC002000.1 - 3 - 4 503 3484605 72
          BC019930.1 - 3 - 4 482 5123891 72
          BM507398.1 - 3 - 4 399 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002479.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sigmar1

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          lipid transport
          nervous system development
          G protein-coupled opioid receptor signaling pathway
          regulation of neuron apoptotic process
          G protein-coupled opioid receptor activity
          protein binding
          signaling receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864091 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864088 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864093 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864092 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864094 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864090 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864089 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045812 BC002000.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045812 BC019930.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045812 AK154300.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045812 AK159886.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045812 AF004927.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169778 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169280 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045812 BC002000.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045812 BC019930.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045812 AK154300.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045812 AK159886.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045812 AF004927.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P300726 AF004927.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864088 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169280 - -
          Whole Mouse Genome G4122A A_51_P300726 AF004927.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127510 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169280 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03633280 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03921011 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03633275 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03633282 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03633276 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864092 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864093 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864091 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864094 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864089 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864088 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03868486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03633274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03358720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03633276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03358702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03633273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03868482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03358704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03293533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03293530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03868495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03293531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03358698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03928208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03633271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03633275 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512314 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512311 - -
          Affymetrix HT MG-430 PM Array Plate 1416750_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1451956_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1440792_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10512308 - -
          Mouse Genome 430 2.0 Array 1416750_at - -
          Mouse Genome 430 2.0 Array 1440792_x_at - -
          Mouse Genome 430 2.0 Array 1440792_PM_x_at - -
          Mouse Genome 430 2.0 Array 1451956_PM_a_at - -
          Mouse Genome 430 2.0 Array 1416750_PM_at - -
          Mouse Genome 430 2.0 Array 1451956_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18391_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18391_at - -
          Affymetrix Mouse Clariom S Array PSR0400020034.mm.1 TC0400002479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020034.mm.1 NM_001286539.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020034.mm.1 NM_001286541.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020034.mm.1 NM_001286542.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020034.mm.1 NM_001286551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020034.mm.1 NM_011014.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020030.mm.1 TC0400002479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020030.mm.1 NM_001286538.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020030.mm.1 NM_001286539.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020030.mm.1 NM_001286540.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020030.mm.1 NM_001286541.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020030.mm.1 NM_001286542.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020030.mm.1 NM_001286551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020030.mm.1 NM_001286605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020030.mm.1 NM_011014.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020037.mm.1 TC0400002479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020037.mm.1 NM_001286538.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020037.mm.1 NM_001286540.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020037.mm.1 NM_001286541.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020037.mm.1 NM_001286551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020037.mm.1 NM_001286605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020037.mm.1 NM_011014.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020033.mm.1 TC0400002479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020033.mm.1 NM_001286539.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020033.mm.1 NM_001286541.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020033.mm.1 NM_001286542.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020033.mm.1 NM_001286551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020033.mm.1 NM_011014.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020031.mm.1 TC0400002479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020031.mm.1 NM_001286538.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020031.mm.1 NM_001286539.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020031.mm.1 NM_001286540.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020031.mm.1 NM_001286541.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020031.mm.1 NM_001286542.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020031.mm.1 NM_001286551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020031.mm.1 NM_001286605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020031.mm.1 NM_011014.3 Refseq
          Mouse Expression Set 430 1416750_at - -
          Mouse Expression Set 430 1451956_a_at - -
          Mouse Expression Set 430 1451956_PM_a_at - -
          Mouse Expression Set 430 1416750_PM_at - -
          Mouse Expression Set 430 1440792_x_at - -
          Mouse Expression Set 430 1440792_PM_x_at - -
          Murine Genome U74v2 Set 94828_at - -
          Murine Genome U74v2 Set 94828_PM_at - -
          Mouse Genome 430A 2.0 Array 1451956_a_at - -
          Mouse Genome 430A 2.0 Array 1416750_at - -
          Mouse Genome 430A 2.0 Array 1416750_PM_at - -
          Mouse Genome 430A 2.0 Array 1451956_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451956_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416750_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451956_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416750_at - -
          Affymetrix HT Mouse Genome 430B Array 1440792_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1440792_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6130128 - -
          Illumina mouse-6 v1.0 expression beadchip 6290717 - -
          Illumina mouse-6 v1.0 expression beadchip 6130128 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006290717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006130128 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238081 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2677170 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2677170 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238081 - -
          mouse-6 v1.1 (Illumina) scl018391.1_236-S - -
          mouse-6 v1.1 (Illumina) scl0002696.1_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 6130128 - -
          Illumina Sentrix mouse-6.v1 bead array 6290717 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl018391.1_236-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002696.1_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290717 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130128 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2677170 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1238081 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2796651 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2677170 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238081 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2796651 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl018391.1_236-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0002696.1_5-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0