Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00449453_m1
          See other USP18 GE Assays ›
          Gene Symbol:
          Usp18
          Gene Name:
          ubiquitin specific peptidase 18
          Gene Aliases:
          1110058H21Rik, AW047653, UBP43, Ubp15
          Chromosome Location:
          Chr.6: 121245906 - 121270917 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00449453_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Usp18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011909.2 NP_036039.2 3 - 4 508 109
          GenBank mRNA AF069502.1 - 3 - 4 469 109
          AK004296.1 - 3 - 4 478 109
          AK149772.1 - 3 - 4 509 109
          AK151011.1 - 3 - 4 509 109
          AK151260.1 - 3 - 4 509 109
          AK151722.1 - 3 - 4 509 109
          AK152034.1 - 3 - 4 474 109
          AK152574.1 - 3 - 4 474 109
          AK153312.1 - 3 - 4 478 109
          BC138562.1 - 3 - 4 390 8861582 109
          BC138577.1 - 3 - 4 390 8861597 109
          CT010228.1 - 2 - 3 244 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001369.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Usp18

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          ubiquitin-dependent protein catabolic process
          protein deubiquitination
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of proteasomal protein catabolic process
          cysteine-type endopeptidase activity
          thiol-dependent deubiquitinase
          protein binding
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity
          pyroglutamyl-peptidase activity
          ISG15-specific protease activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600010888.mm.1 TC0600001369.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600010888.mm.1 NM_011909.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010888.mm.1 NM_011909.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010884.mm.1 TC0600001369.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600010884.mm.1 NM_011909.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010884.mm.1 NM_011909.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010885.mm.1 TC0600001369.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600010885.mm.1 NM_011909.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010885.mm.1 NM_011909.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010886.mm.1 TC0600001369.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600010886.mm.1 NM_011909.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010886.mm.1 NM_011909.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010887.mm.1 TC0600001369.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600010887.mm.1 NM_011909.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010887.mm.1 NM_011909.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 446807 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0