Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00449726_m1
          See other SOD2 GE Assays ›
          Gene Symbol:
          Sod2
          Gene Name:
          superoxide dismutase 2, mitochondrial
          Gene Aliases:
          MnSOD, Sod-2
          Chromosome Location:
          Chr.17: 13007839 - 13018119 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00449726_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sod2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013671.3 NP_038699.2 2 - 3 433 67
          GenBank mRNA AK002428.1 - 2 - 3 293 67
          AK002534.1 - 2 - 3 294 67
          AK012354.1 - 2 - 3 291 67
          AK135435.1 - 2 - 3 422 67
          AK152047.1 - 2 - 3 291 67
          AK168082.1 - 2 - 3 294 67
          AK168591.1 - 2 - 3 425 67
          AK179113.1 - 1 - 2 53 67
          AK180974.1 - 1 - 2 53 67
          BC010548.1 - 2 - 3 269 3581909 67
          BC018173.1 - 1 - 2 214 67
          CT010273.1 - 2 - 3 226 67
          X04972.1 - 2 - 3 278 67
          Z18857.1 - 2 - 3 261 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000148.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Mouse Alzheimer's Disease Inventoried Mouse_Alzheimer Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sod2

          Panther Classification:

          Molecular Function -

          oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          response to superoxide
          age-dependent response to oxidative stress
          age-dependent response to reactive oxygen species
          release of cytochrome c from mitochondria
          liver development
          detection of oxygen
          vasodilation by acetylcholine involved in regulation of systemic arterial blood pressure
          regulation of transcription from RNA polymerase II promoter
          glutathione metabolic process
          superoxide metabolic process
          response to oxidative stress
          mitochondrion organization
          heart development
          sensory perception of sound
          locomotory behavior
          regulation of blood pressure
          negative regulation of cell proliferation
          intrinsic apoptotic signaling pathway in response to DNA damage
          intrinsic apoptotic signaling pathway in response to oxidative stress
          apoptotic mitochondrial changes
          post-embryonic development
          response to gamma radiation
          response to activity
          removal of superoxide radicals
          respiratory electron transport chain
          hemopoiesis
          response to nutrient levels
          oxygen homeostasis
          vasodilation
          response to drug
          response to hydrogen peroxide
          superoxide anion generation
          hydrogen peroxide metabolic process
          negative regulation of apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of nitric oxide biosynthetic process
          negative regulation of fat cell differentiation
          negative regulation of fibroblast proliferation
          neuron development
          response to axon injury
          erythrophore differentiation
          hydrogen peroxide biosynthetic process
          regulation of catalytic activity
          protein homooligomerization
          protein homotetramerization
          regulation of mitochondrial membrane potential
          iron ion homeostasis
          response to hyperoxia
          oxidation-reduction process
          negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          DNA binding
          superoxide dismutase activity
          protein binding
          oxidoreductase activity
          oxygen binding
          manganese ion binding
          identical protein binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700001137.mm.1 NM_013671.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001137.mm.1 NM_013671.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001137.mm.1 TC1700000148.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-z8v7b:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0