Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3kbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135727.2 | NP_001129199.1 | 13 - 14 | 1799 | 82 | |
NM_001135728.2 | NP_001129200.1 | 8 - 9 | 801 | 82 | ||
NM_001290661.1 | NP_001277590.1 | 2 - 3 | 481 | 82 | ||
NM_001290664.1 | NP_001277593.1 | 2 - 3 | 481 | 82 | ||
NM_021389.6 | NP_067364.2 | 12 - 13 | 1364 | 82 | ||
NR_110970.1 | 12 - 13 | 1360 | 82 | |||
GenBank mRNA | AK004636.1 | - | 12 - 13 | 1364 | 82 | |
AK007283.1 | - | 2 - 3 | 468 | 82 | ||
AK028299.1 | - | 8 - 9 | 805 | 82 | ||
AK049182.1 | - | 13 - 14 | 1800 | 82 | ||
AK052260.1 | - | 2 - 3 | 156 | 82 | ||
AK080930.1 | - | 13 - 14 | 1664 | 82 | ||
AK142295.1 | - | 2 - 3 | 255 | 82 | ||
AK161512.1 | - | 2 - 3 | 482 | 82 | ||
AK161735.1 | - | 8 - 9 | 882 | 82 | ||
AK172022.1 | - | 13 - 14 | 1780 | 82 | ||
AK208632.1 | - | 2 - 3 | 173 | 82 | ||
BC034646.1 | - | 12 - 13 | 1369 | 82 | ||
BC052863.1 | - | 12 - 13 | 1309 | 82 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |