Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00452264_m1
          See other RPS19 GE Assays ›
          Gene Symbol:
          Rps19
          Gene Name:
          ribosomal protein S19
          Gene Aliases:
          Dsk3
          Chromosome Location:
          Chr.7: 24884376 - 24889806 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00452264_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023133.1 NP_075622.1 1 - 2 161 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000401.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_023133.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps19

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal small subunit assembly
          maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          monocyte chemotaxis
          rRNA processing
          translation
          nucleolus organization
          Notch signaling pathway
          erythrocyte differentiation
          maturation of SSU-rRNA
          ribosomal small subunit biogenesis
          protein tetramerization
          positive regulation of respiratory burst involved in inflammatory response
          negative regulation of respiratory burst involved in inflammatory response
          RNA binding
          structural constituent of ribosome
          fibroblast growth factor binding
          protein kinase binding
          translation initiation factor binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007072 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007070 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007068 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007067 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007074 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007071 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098471 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38039 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151065 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136849 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098471 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337290 NM_023133.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P404955 AK012052.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P129131 AK031403.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007061 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136849 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06142045 - -
          Whole Mouse Genome G4122A A_52_P404955 AK012052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337290 NM_023133.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P129131 AK031403.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136849 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06272193 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06272174 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06272186 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06643189 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007068 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007070 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007072 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007067 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007074 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06491161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06272156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05950878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06491163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06272154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06272186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06491162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05893531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06643189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06272153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06272184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06272162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06272196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05950881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007113 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551022 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414875 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551023 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551024 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414768 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398600 - -
          Affymetrix HT MG-430 PM Array Plate 1449243_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10414767 - -
          Affymetrix Mouse Gene 1.1 ST Array 10398599 - -
          Affymetrix Mouse Gene 1.1 ST Array 10551021 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414874 - -
          Mouse Genome 430 2.0 Array 1449243_PM_a_at - -
          Mouse Genome 430 2.0 Array 1449243_a_at - -
          Mouse Genome 430 2.0 Array 1460442_PM_at - -
          Mouse Genome 430 2.0 Array 1460442_at - -
          Affymetrix Mouse Clariom S Array PSR0700002978.mm.1 TC0700000401.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002978.mm.1 NM_023133.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002978.mm.1 NM_023133.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002983.mm.1 TC0700000401.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002983.mm.1 NM_023133.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002983.mm.1 NM_023133.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002990.mm.1 TC0700000401.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002990.mm.1 NM_023133.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002990.mm.1 NM_023133.1 Refseq
          Mouse Expression Set 430 1449243_PM_a_at - -
          Mouse Expression Set 430 1460442_PM_at - -
          Mouse Expression Set 430 1460442_at - -
          Mouse Expression Set 430 1449243_a_at - -
          Murine Genome U74v2 Set 94068_at - -
          Murine Genome U74v2 Set 94068_PM_at - -
          Mouse Genome 430A 2.0 Array 1449243_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1460442_PM_at - -
          Mouse Genome 430A 2.0 Array 1449243_a_at - -
          Mouse Genome 430A 2.0 Array 1460442_at - -
          Affymetrix HT Mouse Genome 430A Array 1449243_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449243_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 892239 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5860066 - -
          Illumina mouse-6 v1.0 expression beadchip 5860066 - -
          Illumina mouse-6 v1.0 expression beadchip 101410100 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005860066 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2754355 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2754355 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2461289 - -
          mouse-6 v1.1 (Illumina) scl0020085.2_0-S - -
          mouse-6 v1.1 (Illumina) scl32945.4_345-S - -
          Illumina Sentrix mouse-6.v1 bead array 5860066 - -
          Illumina Sentrix mouse-6.v1 bead array 101410100 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0020085.2_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101410100 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5860066 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2862280 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2754355 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2862280 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2754355 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0