Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00452479_g1
          See other MRPL38 GE Assays ›
          Gene Symbol:
          Mrpl38
          Gene Name:
          mitochondrial ribosomal protein L38
          Gene Aliases:
          1110036N21Rik, 4733401F03Rik, MRP-L3, R75240, Rpml3
          Chromosome Location:
          Chr.11: 116131817 - 116138868 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00452479_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrpl38
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024177.3 NP_077139.2 8 - 9 1097 63
          GenBank mRNA AK088343.1 - 8 - 9 1037 63
          AK151598.1 - 8 - 9 1036 63
          AK159033.1 - 8 - 9 1098 63
          BC002319.1 - 7 - 8 1073 3593984 63
          BC030739.1 - 8 - 9 1024 1493919 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004104.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrpl38

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28157574 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28157576 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28157577 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28157575 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28157579 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352562 AK159033.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352562 BC002319.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352562 BC030739.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352562 NM_024177.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352562 AK088343.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352562 AK159033.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352562 BC002319.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352562 BC030739.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352562 NM_024177.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352562 AK088343.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 548171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 285200 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 814494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 123359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141564 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 115609 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155807 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146915 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352562 AK159033.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352562 BC002319.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352562 BC030739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352562 NM_024177.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352562 AK088343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352562 AK159033.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352562 BC002319.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352562 BC030739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352562 NM_024177.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352562 AK088343.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157559 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155807 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146915 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02438272 - -
          Whole Mouse Genome G4122A A_52_P548227 AK159033.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P548227 BC002319.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P548227 NM_024177.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P548227 AK088343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P548227 AK151598.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352562 AK159033.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352562 BC002319.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352562 BC030739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352562 NM_024177.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352562 AK088343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352562 AK159033.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352562 BC002319.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352562 BC030739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352562 NM_024177.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352562 AK088343.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113934 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102231 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146915 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155807 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09509085 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09509086 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09509081 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09636718 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09656594 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09636715 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09509082 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09509091 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28157579 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28157574 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28157576 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28157577 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28157575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09170474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09143249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09509087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09509091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09509086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09740498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09509085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09335572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09397313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157555 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10393135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393129 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393128 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393126 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393133 - -
          Affymetrix HT MG-430 PM Array Plate 1430610_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1444217_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1447961_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10393125 - -
          Mouse Genome 430 2.0 Array 1430610_PM_at - -
          Mouse Genome 430 2.0 Array 1447961_s_at - -
          Mouse Genome 430 2.0 Array 1447961_PM_s_at - -
          Mouse Genome 430 2.0 Array 1444217_at - -
          Mouse Genome 430 2.0 Array 1430610_at - -
          Mouse Genome 430 2.0 Array 1444217_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60441_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60441_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100038699.mm.1 TC1100004104.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100038699.mm.1 NM_024177.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038699.mm.1 NM_024177.3 Refseq
          Mouse Expression Set 430 1447961_PM_s_at - -
          Mouse Expression Set 430 1447961_s_at - -
          Mouse Expression Set 430 1444217_at - -
          Mouse Expression Set 430 1444217_PM_at - -
          Murine Genome U74v2 Set 96335_PM_at - -
          Murine Genome U74v2 Set 96335_at - -
          Murine Genome U74v2 Set 170959_r_at - -
          Murine Genome U74v2 Set 170959_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1447961_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1447961_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1430610_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1447961_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1447961_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1430610_at - -
          Affymetrix HT Mouse Genome 430B Array 1444217_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444217_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1940154 - -
          Illumina mouse-6 v1.0 expression beadchip 1940154 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001940154 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2664049 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2664049 - -
          mouse-6 v1.1 (Illumina) scl39316.7.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 1940154 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39316.7.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1940154 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2664049 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2664049 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl39316.7.1_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0