Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009439.1 | NP_033465.1 | 1 - 2 | 381 | 69 | |
GenBank mRNA | AK031366.1 | - | 1 - 2 | 371 | 69 | |
AK077252.1 | - | 1 - 2 | 330 | 69 | ||
AK089933.1 | - | 1 - 2 | 339 | 69 | ||
AK145467.1 | - | 1 - 2 | 371 | 69 | ||
AK145488.1 | - | 1 - 2 | 370 | 69 | ||
AK153505.1 | - | 1 - 2 | 298 | 69 | ||
AK164333.1 | - | 1 - 2 | 345 | 69 | ||
AK164593.1 | - | 1 - 2 | 370 | 69 | ||
AK165520.1 | - | 1 - 2 | 316 | 69 | ||
AK166683.1 | - | 1 - 2 | 341 | 69 | ||
AK166714.1 | - | 1 - 2 | 369 | 69 | ||
AK167005.1 | - | 1 - 2 | 356 | 69 | ||
AK167285.1 | - | 1 - 2 | 336 | 69 | ||
AK169690.1 | - | 1 - 2 | 370 | 69 | ||
BC003197.1 | - | 1 - 2 | 381 | 3586427 | 69 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |