Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00456803_m1
          See other MTCP1 GE Assays ›
          Gene Symbol:
          Mtcp1
          Gene Name:
          mature T cell proliferation 1
          Gene Aliases:
          Cmc4, P13MTCP1, p8MTCP1
          Chromosome Location:
          Chr.X: 75404846 - 75416584 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00456803_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mtcp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001294277.1 NP_001281206.1 3 - 4 314 70
          NM_010839.6 NP_034969.1 2 - 3 638 70
          GenBank mRNA BC138811.1 - 2 - 3 240 8861833 70
          BC138812.1 - 2 - 3 240 8861834 70
          Z35294.1 - 2 - 3 598 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002476.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mtcp1

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32471621 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32471622 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32471625 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972634 Z35294.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972634 NM_010839.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972628 U32332.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972628 NM_001039373.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991763 U32332.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991763 NM_001039373.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648236 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 574081 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407772 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25554 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106946 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134109 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991763 U32332.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991763 NM_001039373.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972628 U32332.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972628 NM_001039373.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972634 Z35294.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972634 NM_010839.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32471617 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106946 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134109 - -
          Whole Mouse Genome G4122A A_51_P452280 BC138811.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P452280 BC138812.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P452280 Z35294.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P452280 NM_010839.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P405974 U32332.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P405974 NM_001039373.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136747 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106946 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134109 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15114484 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14755586 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15114483 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15114479 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32471622 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32471625 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32471621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32471497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14823508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15120578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15180879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14756936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15120577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32471617 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10605424 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605423 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605425 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605422 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605428 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605426 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605427 - -
          Affymetrix HT MG-430 PM Array Plate 1449897_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1455710_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1425755_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456207_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10605421 - -
          Mouse Genome 430 2.0 Array 1449897_PM_a_at - -
          Mouse Genome 430 2.0 Array 1456207_PM_at - -
          Mouse Genome 430 2.0 Array 1425755_at - -
          Mouse Genome 430 2.0 Array 1425755_PM_at - -
          Mouse Genome 430 2.0 Array 1455710_PM_x_at - -
          Mouse Genome 430 2.0 Array 1456207_at - -
          Mouse Genome 430 2.0 Array 1449897_a_at - -
          Mouse Genome 430 2.0 Array 1455710_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17763_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17763_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00015827.mm.1 TC0X00002476.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00015827.mm.1 NM_010839.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015828.mm.1 TC0X00002476.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00015828.mm.1 NM_010839.6 Refseq
          Mouse Expression Set 430 1449897_PM_a_at - -
          Mouse Expression Set 430 1455710_PM_x_at - -
          Mouse Expression Set 430 1455710_x_at - -
          Mouse Expression Set 430 1425755_at - -
          Mouse Expression Set 430 1449897_a_at - -
          Mouse Expression Set 430 1425755_PM_at - -
          Mouse Expression Set 430 1456207_PM_at - -
          Mouse Expression Set 430 1456207_at - -
          Murine Genome U74v2 Set 103044_g_at - -
          Murine Genome U74v2 Set 103045_at - -
          Murine Genome U74v2 Set 103044_PM_g_at - -
          Murine Genome U74v2 Set 103043_PM_at - -
          Murine Genome U74v2 Set 103043_at - -
          Murine Genome U74v2 Set 103045_PM_at - -
          Mouse Genome 430A 2.0 Array 1449897_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1425755_PM_at - -
          Mouse Genome 430A 2.0 Array 1455710_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1449897_a_at - -
          Mouse Genome 430A 2.0 Array 1425755_at - -
          Mouse Genome 430A 2.0 Array 1455710_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1449897_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1455710_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1425755_at - -
          Affymetrix HT Mouse Genome 430A Array 1425755_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455710_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1449897_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1456207_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456207_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 838207 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5270068 - -
          Illumina mouse-6 v1.0 expression beadchip 5270068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005270068 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162682 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234358 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234358 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162682 - -
          mouse-6 v1.1 (Illumina) GI_86439950-I - -
          mouse-6 v1.1 (Illumina) scl54116.7.1_56-S - -
          Illumina Sentrix mouse-6.v1 bead array 5270068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54116.7.1_56-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5270068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234358 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3162682 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3162682 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234358 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0