Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00457952_g1
          See other TNIP1 GE Assays ›
          Gene Symbol:
          Tnip1
          Gene Name:
          TNFAIP3 interacting protein 1
          Gene Aliases:
          ABIN, ABIN-1, ABIN1, AU018810, Naf1, Nef, VAN
          Chromosome Location:
          Chr.11: 54910787 - 54962958 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00457952_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnip1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199275.2 NP_001186204.1 11 - 12 1346 63
          NM_001199276.2 NP_001186205.1 9 - 10 1016 63
          NM_001271455.1 NP_001258384.1 10 - 11 1069 63
          NM_001271456.1 NP_001258385.1 11 - 12 1241 63
          NM_021327.4 NP_067302.2 10 - 11 1188 63
          GenBank mRNA AJ242777.1 - 9 - 10 988 63
          AJ242778.1 - 10 - 11 1160 63
          AK135338.1 - 10 - 11 1167 63
          AK156131.1 - 10 - 11 1042 63
          AK170841.1 - 9 - 10 994 63
          AK172890.1 - 974 63
          AK183621.1 - 1 - 2 33 63
          AY012159.2 - 7 - 8 758 63
          BC008186.1 - 10 - 11 1105 3589311 63
          BC008665.1 - 11 - 12 1212 3598934 63
          CB599739.1 - 1 - 2 75 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002770.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnip1

          Gene Ontology Categories:

          Function(s) Process(es)

          MyD88-dependent toll-like receptor signaling pathway
          inflammatory response
          leukocyte cell-cell adhesion
          glycoprotein biosynthetic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of transcription by RNA polymerase II
          positive regulation of inflammatory response
          negative regulation of ERK1 and ERK2 cascade
          modulation by symbiont of host I-kappaB kinase/NF-kappaB cascade
          thiol-dependent deubiquitinase
          protein binding
          polyubiquitin modification-dependent protein binding
          mitogen-activated protein kinase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100025104.mm.1 TC1100002770.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025104.mm.1 NM_001199275.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025104.mm.1 NM_001199276.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025104.mm.1 NM_001271455.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025104.mm.1 NM_001271456.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025104.mm.1 NM_021327.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025105.mm.1 TC1100002770.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025105.mm.1 NM_001199275.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025105.mm.1 NM_001199276.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025105.mm.1 NM_001271455.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025105.mm.1 NM_001271456.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025105.mm.1 NM_021327.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0