Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00459975_mH
          See other CD209D GE Assays ›
          Gene Symbol:
          Cd209d
          Gene Name:
          CD209d antigen
          Gene Aliases:
          SIGN-R3, SIGNR3, mSIGNR3
          Chromosome Location:
          Chr.8: 3871824 - 3878548 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00459975_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd209d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130904.2 NP_570974.1 6 - 7 572 71
          GenBank mRNA AF373411.1 - 6 - 7 572 71
          AF440280.1 - 5 - 6 478 71
          AK136586.1 - 6 - 7 628 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001623.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd209d

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cytokine production
          endocytosis
          defense response to bacterium
          mannose binding
          carbohydrate binding
          protein homodimerization activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2055985 NM_130904.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2055985 AF373411.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2055985 AK136586.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 15189 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921602 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157020 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2055985 NM_130904.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2055985 AF373411.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2055985 AK136586.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615689 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06408348 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157020 - -
          Whole Mouse Genome G4122A A_52_P401535 NM_130904.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P401535 AF373411.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P401535 AF440280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P401535 AK136586.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123517 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157020 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07254819 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06761025 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07254816 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07254820 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07254812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06761025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06743525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06684577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06797756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06797754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06797755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07248917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07365303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06743514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06684580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615672 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10576809 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576811 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576813 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576810 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576812 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576808 - -
          Affymetrix HT MG-430 PM Array Plate 1426183_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10576807 - -
          Mouse Genome 430 2.0 Array 1426183_PM_a_at - -
          Mouse Genome 430 2.0 Array 1426183_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170779_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170779_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800013343.mm.1 TC0800001623.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013343.mm.1 NM_130904.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013343.mm.1 NM_130904.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013345.mm.1 TC0800001623.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013345.mm.1 NM_130904.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013345.mm.1 NM_130904.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013344.mm.1 TC0800001623.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013344.mm.1 NM_130904.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013344.mm.1 NM_130904.2 Refseq
          Mouse Expression Set 430 1426183_a_at - -
          Mouse Expression Set 430 1426183_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1426183_a_at - -
          Mouse Genome 430A 2.0 Array 1426183_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1426183_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1426183_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 812277 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2470593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002470593 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2696291 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2696291 - -
          mouse-6 v1.1 (Illumina) scl35149.7.1_82-S - -
          Illumina Sentrix mouse-6.v1 bead array 2470593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35149.7.1_82-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2470593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2696291 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2696291 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0