Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00461022_m1
          See other JMJD7 GE Assays ›
          Gene Symbol:
          Jmjd7 Gm28042
          Gene Name:
          jumonji domain containing 7
          predicted gene, 28042
          Gene Aliases:
          -
          Jmjd7-pla2g4b
          Chromosome Location:
          Chr.2: 120027483 - 120032604 on Build GRCm38
          Chromosome Location:
          Chr.2: 120027483 - 120043033 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00461022_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Jmjd7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001114637.1 NP_001108109.1 1 - 2 97 70
          GenBank mRNA BC016255.1 - 1 - 2 74 70
          Gene Symbol: Gm28042
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_104353.1 1 - 2 97 70
          GenBank mRNA AB195274.1 - 1 - 2 87 70
          AB195275.1 - 1 - 2 87 70

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AB195274.1, AB195275.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P288549 BC016255.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P288549 BC016255.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810417 AK086670.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810417 BC031513.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810417 BC066060.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810417 BC066087.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810417 NM_153600.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 910563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140608 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171522 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P288549 BC016255.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P288549 BC016255.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P288549 BC016255.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P288549 BC016255.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21572838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01755268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02172844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01707444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01755273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01309532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21572788 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10475120 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475123 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475119 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475118 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475122 - -
          Affymetrix Mouse Gene 1.1 ST Array 10475113 - -
          Mouse Genome 430 2.0 Array 1425045_at - -
          Mouse Genome 430 2.0 Array 1425045_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200013545.mm.1 TC0200001788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013545.mm.1 NM_001114637.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013545.mm.1 NM_001114637.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013545.mm.1 NR_104353.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013550.mm.1 TC0200001788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013550.mm.1 NM_001114637.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013550.mm.1 NM_001114637.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013550.mm.1 NR_104353.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013552.mm.1 TC0200001788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013552.mm.1 NM_001114637.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013552.mm.1 NM_001114637.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013552.mm.1 NR_104353.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013565.mm.1 TC0200001788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013565.mm.1 NM_001114637.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013565.mm.1 NM_001114637.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013549.mm.1 TC0200001788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013549.mm.1 NM_001114637.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013549.mm.1 NM_001114637.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013549.mm.1 NR_104353.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013551.mm.1 TC0200001788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013551.mm.1 NM_001114637.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013551.mm.1 NM_001114637.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013551.mm.1 NR_104353.1 Refseq
          Mouse Expression Set 430 1425045_PM_at - -
          Mouse Expression Set 430 1425045_at - -
          Mouse Expression Set 430 1456047_PM_at - -
          Mouse Expression Set 430 1456047_at - -
          Murine Genome U74v2 Set 168171_PM_s_at - -
          Murine Genome U74v2 Set 166864_PM_at - -
          Murine Genome U74v2 Set 168171_s_at - -
          Murine Genome U74v2 Set 166864_at - -
          Mouse Genome 430A 2.0 Array 1425045_at - -
          Mouse Genome 430A 2.0 Array 1425045_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0