Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00461839_m1
          See other MKL1 GE Assays ›
          Gene Symbol:
          Mkl1
          Gene Name:
          MKL (megakaryoblastic leukemia)/myocardin-like 1
          Gene Aliases:
          AMKL, Bsac, Mal, Mrtf-A
          Chromosome Location:
          Chr.15: 81012281 - 81190757 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00461839_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mkl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001082536.1 NP_001076005.1 3 - 4 578 80
          NM_153049.3 NP_694629.2 2 - 3 340 80
          GenBank mRNA AF385582.1 - 2 - 3 111 80
          AF532597.1 - 3 - 4 462 80
          AK044188.1 - 3 - 4 571 80
          AK089416.1 - 1 - 2 170 80
          AK154561.1 - 3 - 4 558 80
          AK155821.1 - 3 - 4 580 80
          AK155937.1 - 2 - 3 324 80
          AK157985.1 - 1 - 2 189 80
          BB850443.1 - 2 - 3 323 80
          BC050941.1 - 3 - 4 471 6821370 80
          BC054801.1 - 3 - 4 514 80
          BY200528.1 - 2 - 3 313 80
          BY208699.1 - 2 - 3 341 80
          BY749697.1 - 1 - 2 170 80

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF532597.1, AK044188.1, AK089416.1, AK154561.1, BC050941.1, NM_001082536.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mkl1

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          positive regulation of transcription via serum response element binding
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          smooth muscle cell differentiation
          negative regulation of apoptotic signaling pathway
          transcription cis-regulatory region binding
          DNA-binding transcription factor activity
          transcription coactivator activity
          actin binding
          actin monomer binding
          protein binding
          leucine zipper domain binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500015029.mm.1 TC1500001912.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500015029.mm.1 NM_001082536.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015029.mm.1 NM_001082536.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015019.mm.1 TC1500001912.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500015019.mm.1 NM_001082536.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015019.mm.1 NM_001082536.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015019.mm.1 NM_153049.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015022.mm.1 TC1500001912.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500015022.mm.1 NM_153049.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015025.mm.1 TC1500001912.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500015025.mm.1 NM_001082536.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015025.mm.1 NM_001082536.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015023.mm.1 TC1500001912.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500015023.mm.1 NM_153049.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015028.mm.1 TC1500001912.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500015028.mm.1 NM_001082536.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015028.mm.1 NM_001082536.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015017.mm.1 TC1500001912.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500015017.mm.1 NM_001082536.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015017.mm.1 NM_001082536.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015017.mm.1 NM_153049.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 744446 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0