Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200009406.mm.1 |
TC1200001302.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009406.mm.1 |
NM_028731.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009406.mm.1 |
NM_028731.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009403.mm.1 |
TC1200001302.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009403.mm.1 |
NM_028731.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009403.mm.1 |
NM_028731.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009399.mm.1 |
TC1200001302.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009399.mm.1 |
NM_028731.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009399.mm.1 |
NM_028731.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009401.mm.1 |
TC1200001302.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009401.mm.1 |
NM_028731.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009401.mm.1 |
NM_028731.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009402.mm.1 |
TC1200001302.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009402.mm.1 |
NM_028731.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009402.mm.1 |
NM_028731.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009405.mm.1 |
TC1200001302.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009405.mm.1 |
NM_028731.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009405.mm.1 |
NM_028731.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009400.mm.1 |
TC1200001302.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009400.mm.1 |
NM_028731.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009400.mm.1 |
NM_028731.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009404.mm.1 |
TC1200001302.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009404.mm.1 |
NM_028731.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009404.mm.1 |
NM_028731.5 |
Refseq |