Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00468038_m1
          See other SPATA2 GE Assays ›
          Gene Symbol:
          Spata2
          Gene Name:
          spermatogenesis associated 2
          Gene Aliases:
          AI504642, mKIAA0757
          Chromosome Location:
          Chr.2: 167481136 - 167492874 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00468038_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spata2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_170756.2 NP_739562.1 1 - 2 117 59
          GenBank mRNA AK051741.1 - 1 - 2 63 59
          AK144826.1 - 1 - 2 89 59
          AK156188.1 - 1 - 2 118 59
          AK162743.1 - 1 - 2 117 59
          AK170356.1 - 1 - 2 118 59
          AK171240.1 - 1 - 2 91 59
          AK171664.1 - 1 - 2 95 59
          AK171852.1 - 1 - 2 107 59
          BC034597.1 - 1 - 2 106 5363513 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005189.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK051741.1, AK144826.1, AK156188.1, AK162743.1, AK170356.1, AK171664.1, AK171852.1, BC034597.1, NM_170756.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spata2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21835810 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21835811 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21835809 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21835808 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152605 BC034597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152605 AK122364.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152605 AK171664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152605 AK170356.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152605 AK171240.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147141 BC034597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147141 AK122364.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147141 AK171852.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147141 AK171664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147141 AK144826.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P514306 BC034597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P514306 AK122364.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P514306 AK171852.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P514306 AK171664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P514306 AK156188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P514306 BC034597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P514306 AK122364.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P514306 AK171852.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P514306 AK171664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P514306 AK156188.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 642421 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825168 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 273533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 484228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865975 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319493 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51775 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140367 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173665 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P514306 BC034597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P514306 AK122364.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P514306 AK171852.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P514306 AK171664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P514306 AK156188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P514306 BC034597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P514306 AK122364.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P514306 AK171852.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P514306 AK171664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P514306 AK156188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147141 BC034597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147141 AK122364.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147141 AK171852.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147141 AK171664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147141 AK144826.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152605 BC034597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152605 AK122364.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152605 AK171664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152605 AK170356.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152605 AK171240.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21835826 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140367 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173665 - -
          Whole Mouse Genome G4122A A_51_P110699 BC034597.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P110699 AK122364.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P110699 AK171852.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P110699 AK171664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P110699 AK170356.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P514306 BC034597.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P514306 AK122364.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P514306 AK171852.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P514306 AK171664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P514306 AK156188.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P514306 BC034597.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P514306 AK122364.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P514306 AK171852.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P514306 AK171664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P514306 AK156188.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173665 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140367 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01655100 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01719081 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01655095 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01655097 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01996287 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21835809 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21835810 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21835811 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21835808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21835825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01560127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01649491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01208368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02129875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02152371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01996287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01310910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02129872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01165671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01165673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21835825 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10489906 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489908 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489907 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489905 - -
          Affymetrix HT MG-430 PM Array Plate 1456776_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434107_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10489904 - -
          Mouse Genome 430 2.0 Array 1434107_at - -
          Mouse Genome 430 2.0 Array 1434107_PM_at - -
          Mouse Genome 430 2.0 Array 1456776_PM_at - -
          Mouse Genome 430 2.0 Array 1456776_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 263876_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 263876_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200044434.mm.1 TC0200005189.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200044434.mm.1 NM_170756.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044434.mm.1 NM_170756.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044435.mm.1 TC0200005189.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200044435.mm.1 NM_170756.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044435.mm.1 NM_170756.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044444.mm.1 TC0200005189.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200044444.mm.1 NM_170756.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044444.mm.1 NM_170756.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044443.mm.1 TC0200005189.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200044443.mm.1 NM_170756.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044443.mm.1 NM_170756.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044436.mm.1 TC0200005189.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200044436.mm.1 NM_170756.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044436.mm.1 NM_170756.2 Refseq
          Mouse Expression Set 430 1456776_PM_at - -
          Mouse Expression Set 430 1434107_PM_at - -
          Mouse Expression Set 430 1434107_at - -
          Mouse Expression Set 430 1456776_at - -
          Affymetrix HT Mouse Genome 430B Array 1434107_at - -
          Affymetrix HT Mouse Genome 430B Array 1456776_at - -
          Affymetrix HT Mouse Genome 430B Array 1434107_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456776_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 529710 - -
          ABI Mouse Genome Survey Microarray 533140 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5910541 - -
          MouseWG-6 v2 Expression BeadChip 1230215 - -
          Illumina mouse-6 v1.0 expression beadchip 5910541 - -
          Illumina mouse-6 v1.0 expression beadchip 1230215 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005910541 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001230215 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2640415 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2692815 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2692815 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2640415 - -
          mouse-6 v1.1 (Illumina) scl00263876.2_276-S - -
          mouse-6 v1.1 (Illumina) scl00263876.2_86-S - -
          Illumina Sentrix mouse-6.v1 bead array 5910541 - -
          Illumina Sentrix mouse-6.v1 bead array 1230215 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00263876.2_276-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00263876.2_86-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5910541 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1230215 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2692815 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2640415 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2640415 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2692815 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-ffb8c786-92mpb:80/100.66.129.117:80.
          git-commit: 1a801765356e65be0b6ed8595ed79b52bb698fed
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0