Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00468761_m1
          See other ERF GE Assays ›
          Gene Symbol:
          Erf Col1a1
          Gene Name:
          Ets2 repressor factor
          collagen, type I, alpha 1
          Gene Aliases:
          -
          Col1a-1, Cola-1, Cola1, Mov-13, Mov13
          Chromosome Location:
          Chr.7: 25242560 - 25250758 on Build GRCm38
          Chromosome Location:
          Chr.11: 94936270 - 94951867 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00468761_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Erf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010155.3 NP_034285.3 1 - 2 183 65
          GenBank mRNA AK159225.1 - 1 - 2 160 65
          BC053045.1 - 1 - 2 139 65
          BC059176.1 - 1 - 2 139 65
          BC063092.1 - 1 - 2 90 30148108 65
          Gene Symbol: Col1a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC042642.1 - 96 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002628.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK159225.1, BC063092.1, NM_010155.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Erf
          Click here to find siRNAs for Gene Col1a1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          in utero embryonic development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          ectodermal cell differentiation
          trophoblast giant cell differentiation
          chorio-allantoic fusion
          DNA-binding transcription factor activity, RNA polymerase II-specific
          DNA binding
          DNA-binding transcription factor activity
          transcription corepressor activity
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P326354 BC059176.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P326354 BC063092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P326354 BC053045.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P326354 NM_010155.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P326354 BC059176.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P326354 BC063092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P326354 BC053045.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P326354 NM_010155.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590210 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142175 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P326354 BC059176.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P326354 BC063092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P326354 BC053045.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P326354 NM_010155.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P326354 BC059176.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P326354 BC063092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P326354 BC053045.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P326354 NM_010155.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25009240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25009242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25009271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25009273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25009268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25009244 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150763 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06143079 - -
          Whole Mouse Genome G4122A A_52_P326354 BC059176.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P326354 BC063092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P326354 BC053045.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P326354 NM_010155.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P326354 BC059176.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P326354 BC063092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P326354 BC053045.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P326354 NM_010155.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133767 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135423 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150763 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06614361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25009267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06027370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06211890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06027369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06353512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06211883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25009263 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10561006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561005 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561007 - -
          Affymetrix HT MG-430 PM Array Plate 1422114_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435561_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10561004 - -
          Mouse Genome 430 2.0 Array 1422114_at - -
          Mouse Genome 430 2.0 Array 1422114_PM_at - -
          Mouse Genome 430 2.0 Array 1435561_PM_at - -
          Mouse Genome 430 2.0 Array 1435561_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13875_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13875_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700022707.mm.1 TC0700002628.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700022707.mm.1 NM_010155.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022707.mm.1 NM_010155.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022705.mm.1 TC0700002628.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700022705.mm.1 NM_010155.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022705.mm.1 NM_010155.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022706.mm.1 TC0700002628.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700022706.mm.1 NM_010155.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022706.mm.1 NM_010155.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022708.mm.1 TC0700002628.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700022708.mm.1 NM_010155.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022708.mm.1 NM_010155.3 Refseq
          Mouse Expression Set 430 1422114_at - -
          Mouse Expression Set 430 1422114_PM_at - -
          Mouse Expression Set 430 1435561_PM_at - -
          Mouse Expression Set 430 1435561_at - -
          Murine Genome U74v2 Set 93670_at - -
          Murine Genome U74v2 Set 93671_PM_at - -
          Murine Genome U74v2 Set 93670_PM_at - -
          Murine Genome U74v2 Set 93671_at - -
          Mouse Genome 430A 2.0 Array 1435561_PM_at - -
          Mouse Genome 430A 2.0 Array 1422114_at - -
          Mouse Genome 430A 2.0 Array 1422114_PM_at - -
          Mouse Genome 430A 2.0 Array 1435561_at - -
          Affymetrix HT Mouse Genome 430A Array 1422114_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422114_at - -
          Affymetrix HT Mouse Genome 430A Array 1435561_at - -
          Affymetrix HT Mouse Genome 430A Array 1435561_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 368785 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 110551 - -
          Illumina mouse-6 v1.0 expression beadchip 1770278 - -
          Illumina mouse-6 v1.0 expression beadchip 110551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000110551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228355 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2684661 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228355 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2684661 - -
          mouse-6 v1.1 (Illumina) scl013875.1_31-S - -
          mouse-6 v1.1 (Illumina) scl0013875.2_59-S - -
          Illumina Sentrix mouse-6.v1 bead array 110551 - -
          Illumina Sentrix mouse-6.v1 bead array 1770278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013875.1_31-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0013875.2_59-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770278 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 110551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2684661 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2651005 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2650998 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2684661 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2651005 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2650998 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0