Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100024578.mm.1 |
TC1100002694.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100024578.mm.1 |
NM_001048061.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024578.mm.1 |
NM_001048061.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024578.mm.1 |
NM_010448.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024578.mm.1 |
NM_010448.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024579.mm.1 |
TC1100002694.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100024579.mm.1 |
NM_001048061.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024579.mm.1 |
NM_001048061.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024579.mm.1 |
NM_010448.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024579.mm.1 |
NM_010448.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024581.mm.1 |
TC1100002694.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100024581.mm.1 |
NM_001048061.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024581.mm.1 |
NM_001048061.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024581.mm.1 |
NM_010448.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024581.mm.1 |
NM_010448.3 |
Refseq |