Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00469734_m1
          See other BRD3 GE Assays ›
          Gene Symbol:
          Brd3
          Gene Name:
          bromodomain containing 3
          Gene Aliases:
          2410084F24Rik, AW060456, Fsrg2, ORFX, RINGL3
          Chromosome Location:
          Chr.2: 27445579 - 27507657 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00469734_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Brd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113573.1 NP_001107045.1 10 - 11 2134 89
          NM_001113574.1 NP_001107046.1 11 - 12 2185 89
          NM_023336.4 NP_075825.3 10 - 11 2294 89
          GenBank mRNA AB206708.2 - 10 - 11 1996 89
          AF269193.1 - 10 - 11 2082 89
          AK014429.1 - 2 - 3 346 89
          AK136429.1 - 10 - 11 2066 89
          AK159827.1 - 10 - 11 2120 89
          AK160752.1 - 10 - 11 2093 89
          AK161164.1 - 9 - 10 89
          AK167573.1 - 10 - 11 2092 89
          AY513269.1 - 9 - 10 1960 89
          BC031536.1 - 10 - 11 2108 4015879 89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AK142230.1 - Chr. - - [-] on Build 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003217.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Brd3

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          chromatin binding
          lysine-acetylated histone binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200027146.mm.1 TC0200003217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027146.mm.1 NM_001113573.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027146.mm.1 NM_001113573.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027146.mm.1 NM_001113574.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027146.mm.1 NM_001113574.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027146.mm.1 NM_023336.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027146.mm.1 NM_023336.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027147.mm.1 TC0200003217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027147.mm.1 NM_001113573.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027147.mm.1 NM_001113573.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027147.mm.1 NM_001113574.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027147.mm.1 NM_001113574.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027147.mm.1 NM_023336.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027147.mm.1 NM_023336.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0