Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00470185_m1
          See other PAQR4 GE Assays ›
          Gene Symbol:
          Paqr4
          Gene Name:
          progestin and adipoQ receptor family member IV
          Gene Aliases:
          1500004C10Rik
          Chromosome Location:
          Chr.17: 23736186 - 23740898 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00470185_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Paqr4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023824.3 NP_076313.1 1 - 2 474 62
          GenBank mRNA AB041564.1 - 1 - 2 395 62
          AK005142.1 - 1 - 2 497 62
          AK033625.1 - 1 - 2 492 62
          AK044092.1 - 1 - 2 294 62
          AK044486.1 - 1 - 2 497 62
          AK152676.1 - 1 - 2 473 62
          AY424293.1 - 1 - 2 425 62
          BC011185.1 - 1 - 2 474 4196033 62
          BC026921.1 - 1 - 2 425 4500871 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001602.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Paqr4

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          signaling receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093360 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093354 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093359 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093356 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093353 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093355 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093358 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093357 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109050 BC011185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109050 BC026921.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109050 AK044486.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109050 AK152676.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109050 AY424293.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109050 BC011185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109050 BC026921.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109050 AK044486.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109050 AK152676.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109050 AY424293.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478677 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25841 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138610 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113541 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109050 BC011185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109050 BC026921.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109050 AK044486.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109050 AK152676.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109050 AY424293.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109050 BC011185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109050 BC026921.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109050 AK044486.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109050 AK152676.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109050 AY424293.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093359 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113541 - -
          Whole Mouse Genome G4122A A_51_P109050 BC011185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109050 BC026921.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109050 AK044486.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109050 AK152676.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109050 AY424293.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109050 BC011185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109050 BC026921.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109050 AK044486.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109050 AK152676.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109050 AY424293.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111531 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113541 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13277189 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13277183 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13277168 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093358 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093353 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093354 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093357 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093359 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093355 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093360 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13196604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13196602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13494239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13277182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13196607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13233751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13233752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093345 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448353 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448352 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448354 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448351 - -
          Affymetrix HT MG-430 PM Array Plate 1423101_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10448350 - -
          Affymetrix Mouse Gene 1.1 ST Array 10442331 - -
          Mouse Genome 430 2.0 Array 1423101_at - -
          Mouse Genome 430 2.0 Array 1423101_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76498_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76498_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700014027.mm.1 TC1700001602.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014027.mm.1 NM_023824.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014027.mm.1 NM_023824.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014026.mm.1 TC1700001602.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014026.mm.1 NM_023824.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014026.mm.1 NM_023824.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014028.mm.1 TC1700001602.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014028.mm.1 NM_023824.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014028.mm.1 NM_023824.3 Refseq
          Mouse Expression Set 430 1423101_at - -
          Mouse Expression Set 430 1423101_PM_at - -
          Murine Genome U74v2 Set 170244_PM_at - -
          Murine Genome U74v2 Set 170244_at - -
          Mouse Genome 430A 2.0 Array 1423101_PM_at - -
          Mouse Genome 430A 2.0 Array 1423101_at - -
          Affymetrix HT Mouse Genome 430A Array 1423101_at - -
          Affymetrix HT Mouse Genome 430A Array 1423101_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 342655 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 730736 - -
          Illumina mouse-6 v1.0 expression beadchip 730736 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000730736 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2748229 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251921 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251921 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2748229 - -
          mouse-6 v1.1 (Illumina) scl50233.3_39-S - -
          Illumina Sentrix mouse-6.v1 bead array 730736 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50233.3_39-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 730736 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2748229 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50233.3_39-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0