Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00470391_g1
          See other B3GAT3 GE Assays ›
          Gene Symbol:
          B3gat3
          Gene Name:
          beta-1,3-glucuronyltransferase 3 (glucuronosyltransferase I)
          Gene Aliases:
          2810405M13Rik
          Chromosome Location:
          Chr.19: 8920393 - 8927236 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00470391_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B3gat3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024256.2 NP_077218.1 3 - 4 788 78
          GenBank mRNA AK144402.1 - 3 - 4 807 78
          AK146264.1 - 3 - 4 699 78
          BC002103.1 - 3 - 4 655 3485805 78
          BC004038.1 - 3 - 4 788 3496170 78
          BC012930.1 - 3 - 4 776 4500993 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000166.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene B3gat3

          Gene Ontology Categories:

          Function(s) Process(es)

          glycosaminoglycan biosynthetic process
          heparan sulfate proteoglycan biosynthetic process
          positive regulation of catalytic activity
          chondroitin sulfate proteoglycan biosynthetic process
          dermatan sulfate proteoglycan biosynthetic process
          positive regulation of intracellular protein transport
          galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase activity
          glucuronosyltransferase activity
          transferase activity
          metal ion binding
          protein phosphatase activator activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P506344 BC004038.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P506344 BC012930.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P506344 AK144402.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P506344 AK146264.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P506344 BC002103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P506344 BC004038.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P506344 BC012930.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P506344 AK144402.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P506344 AK146264.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P506344 BC002103.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778257 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657878 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 116097 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133435 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P506344 BC004038.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P506344 BC012930.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P506344 AK144402.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P506344 AK146264.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P506344 BC002103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P506344 BC004038.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P506344 BC012930.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P506344 AK144402.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P506344 AK146264.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P506344 BC002103.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364432 BC004038.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364432 BC012930.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364432 AK144402.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364432 AK146264.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364432 AK178713.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P201315 AK079723.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P201315 AK013004.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P416774 AK079723.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P201315 AK079723.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P201315 AK013004.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P506344 BC004038.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P506344 BC012930.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P506344 AK144402.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P506344 AK146264.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P506344 BC002103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P506344 BC004038.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P506344 BC012930.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P506344 AK144402.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P506344 AK146264.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P506344 BC002103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364432 BC004038.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364432 BC012930.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364432 AK144402.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364432 AK146264.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364432 AK178713.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100251 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14416042 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14352684 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14393034 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461308 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461310 - -
          Affymetrix HT MG-430 PM Array Plate 1416960_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1431219_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461305 - -
          Mouse Genome 430 2.0 Array 1416960_at - -
          Mouse Genome 430 2.0 Array 1431219_PM_at - -
          Mouse Genome 430 2.0 Array 1416960_PM_at - -
          Mouse Genome 430 2.0 Array 1431219_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72727_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72727_at - -
          Affymetrix Mouse Clariom S Array PSR1900002250.mm.1 TC1900000166.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002250.mm.1 NM_024256.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002250.mm.1 NM_024256.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002249.mm.1 TC1900000166.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002249.mm.1 NM_024256.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002249.mm.1 NM_024256.2 Refseq
          Mouse Expression Set 430 1431219_at - -
          Mouse Expression Set 430 1431219_PM_at - -
          Mouse Expression Set 430 1416960_at - -
          Mouse Expression Set 430 1416960_PM_at - -
          Murine Genome U74v2 Set 94407_PM_at - -
          Murine Genome U74v2 Set 94407_at - -
          Mouse Genome 430A 2.0 Array 1416960_PM_at - -
          Mouse Genome 430A 2.0 Array 1431219_PM_at - -
          Mouse Genome 430A 2.0 Array 1416960_at - -
          Mouse Genome 430A 2.0 Array 1431219_at - -
          Affymetrix HT Mouse Genome 430A Array 1431219_at - -
          Affymetrix HT Mouse Genome 430A Array 1416960_at - -
          Affymetrix HT Mouse Genome 430A Array 1416960_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1431219_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101450736 - -
          MouseWG-6 v2 Expression BeadChip 103360097 - -
          MouseWG-6 v2 Expression BeadChip 105220672 - -
          Illumina mouse-6 v1.0 expression beadchip 101450736 - -
          Illumina mouse-6 v1.0 expression beadchip 1580438 - -
          Illumina mouse-6 v1.0 expression beadchip 105220672 - -
          Illumina mouse-6 v1.0 expression beadchip 103360097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001580438 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2729706 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2729706 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234775 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2547059 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255717 - -
          mouse-6 v1.1 (Illumina) scl53399.4.220_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 105220672 - -
          Illumina Sentrix mouse-6.v1 bead array 1580438 - -
          Illumina Sentrix mouse-6.v1 bead array 103360097 - -
          Illumina Sentrix mouse-6.v1 bead array 101450736 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53399.4.220_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101450736 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105220672 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1580438 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103360097 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2729706 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2845045 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221140 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234775 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2572833 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2729706 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2845045 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl53399.4.220_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0