Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471703_s1
          See other 1700120K04RIK GE Assays ›
          Gene Symbol:
          1700120K04Rik
          Gene Name:
          RIKEN cDNA 1700120K04 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 127603903 - 127604445 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00471703_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700120K04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027915.1 1 242 96
          GenBank mRNA AK007016.1 - 1 235 96
          AK007155.1 - 1 235 96
          AK018960.1 - 1 243 96
          AK018969.1 - 1 242 96
          BC049550.1 - 227 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004333.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501622 AK018960.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501622 AK018969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501622 BC049550.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501622 AK007155.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501622 NR_027915.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501622 AK018960.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501622 AK018969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501622 BC049550.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501622 AK007155.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501622 NR_027915.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952469 AK018960.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952469 AK018969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952469 BC049550.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952469 AK007155.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952469 NR_027915.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501622 AK018960.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501622 AK018969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501622 BC049550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501622 AK007155.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501622 NR_027915.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501622 AK018960.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501622 AK018969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501622 BC049550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501622 AK007155.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501622 NR_027915.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568280 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568281 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568279 - -
          Affymetrix HT MG-430 PM Array Plate 1420681_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568278 - -
          Mouse Genome 430 2.0 Array 1420681_at - -
          Mouse Genome 430 2.0 Array 1420681_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68232_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68232_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700036387.mm.1 TC0700004333.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036387.mm.1 NR_027915.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036387.mm.1 NR_027915.1 Refseq
          Mouse Expression Set 430 1420681_at - -
          Mouse Expression Set 430 1420681_PM_at - -
          Murine Genome U74v2 Set 168141_i_at - -
          Murine Genome U74v2 Set 168141_PM_i_at - -
          Murine Genome U74v2 Set 169074_PM_at - -
          Murine Genome U74v2 Set 169074_at - -
          Mouse Genome 430A 2.0 Array 1420681_at - -
          Mouse Genome 430A 2.0 Array 1420681_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420681_at - -
          Affymetrix HT Mouse Genome 430A Array 1420681_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 677177 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 770161 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0