Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00472301_s1
          See other ACTRT1 GE Assays ›
          Gene Symbol:
          Actrt1
          Gene Name:
          actin-related protein T1
          Gene Aliases:
          1700061J02Rik, Arp-T1
          Chromosome Location:
          Chr.X: 46329007 - 46330345 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00472301_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Actrt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028514.3 NP_082790.1 1 1240 139
          GenBank mRNA AK006855.1 - 1 1241 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000418.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Actrt1

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          positive regulation of mitophagy in response to mitochondrial depolarization
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518094 BC116432.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518094 NM_028514.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518094 AK006855.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518094 BC116431.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518094 BC116432.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518094 NM_028514.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518094 AK006855.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518094 BC116431.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147915 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518094 BC116432.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518094 NM_028514.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518094 AK006855.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518094 BC116431.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518094 BC116432.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518094 NM_028514.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518094 AK006855.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518094 BC116431.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32355688 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147915 - -
          Whole Mouse Genome G4122A A_51_P518094 BC116432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518094 NM_028514.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518094 AK006855.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518094 BC116431.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518094 BC116432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518094 NM_028514.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518094 AK006855.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518094 BC116431.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124067 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147915 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14635155 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14635161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32355670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14635152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14635160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32355676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14635167 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10599426 - -
          Affymetrix HT MG-430 PM Array Plate 1449825_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10599425 - -
          Mouse Genome 430 2.0 Array 1449825_PM_at - -
          Mouse Genome 430 2.0 Array 1449825_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73360_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73360_at - -
          Affymetrix Mouse Clariom S Array PSR0X00002760.mm.1 TC0X00000418.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002760.mm.1 NM_028514.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002760.mm.1 NM_028514.3 Refseq
          Mouse Expression Set 430 1449825_PM_at - -
          Mouse Expression Set 430 1449825_at - -
          Murine Genome U74v2 Set 167445_PM_at - -
          Murine Genome U74v2 Set 167445_at - -
          Mouse Genome 430A 2.0 Array 1449825_PM_at - -
          Mouse Genome 430A 2.0 Array 1449825_at - -
          Affymetrix HT Mouse Genome 430A Array 1449825_at - -
          Affymetrix HT Mouse Genome 430A Array 1449825_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 778744 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4480347 - -
          Illumina mouse-6 v1.0 expression beadchip 4480347 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004480347 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2618100 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2618100 - -
          mouse-6 v1.1 (Illumina) scl54955.1.1_139-S - -
          Illumina Sentrix mouse-6.v1 bead array 4480347 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54955.1.1_139-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4480347 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2618100 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2618100 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline