Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00473363_m1
          See other DBR1 GE Assays ›
          Gene Symbol:
          Dbr1
          Gene Name:
          debranching enzyme homolog 1 (S. cerevisiae)
          Gene Aliases:
          AW018415
          Chromosome Location:
          Chr.9: 99575775 - 99584461 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00473363_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dbr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031403.3 NP_113580.2 7 - 8 1072 75
          GenBank mRNA AK050090.1 - 7 - 8 1073 75
          AK090351.1 - 7 - 8 1048 75
          AK139631.1 - 7 - 8 1056 75
          BC006661.1 - 7 - 8 1067 3501390 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001252.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dbr1

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA splicing, via transesterification reactions
          mRNA splicing, via spliceosome
          mRNA processing
          RNA phosphodiester bond hydrolysis, endonucleolytic
          RNA binding
          nuclease activity
          RNA lariat debranching enzyme activity
          hydrolase activity
          hydrolase activity, acting on ester bonds
          phosphoric ester hydrolase activity
          T/G mismatch-specific endonuclease activity
          retroviral 3' processing activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26764949 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26764950 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255841 AK139631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255841 BC006661.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255841 AK050090.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255841 AK077674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255841 NM_031403.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255841 AK139631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255841 BC006661.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255841 AK050090.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255841 AK077674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255841 NM_031403.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 5531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 166632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335584 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330660 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713460 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294939 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227885 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92508 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176235 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255841 AK139631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255841 BC006661.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255841 AK050090.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255841 AK077674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255841 NM_031403.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255841 AK139631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255841 BC006661.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255841 AK050090.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255841 AK077674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255841 NM_031403.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26764963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26764949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26764952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26764947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26764961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26764965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26764958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26764956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26764960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26764953 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176235 - -
          Whole Mouse Genome G4122A A_52_P255841 AK139631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P255841 BC006661.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P255841 AK050090.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P255841 AK077674.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P255841 NM_031403.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P255841 AK139631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P255841 BC006661.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P255841 AK050090.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P255841 AK077674.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P255841 NM_031403.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117030 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176235 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26764949 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26764950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26765063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26764902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26765058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26765051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08130611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26765053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26765044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07964717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07720941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26765046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07629227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08130610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07700496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26764942 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10588126 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588123 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588128 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588129 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588122 - -
          Affymetrix HT MG-430 PM Array Plate 1451641_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10588121 - -
          Mouse Genome 430 2.0 Array 1451641_PM_at - -
          Mouse Genome 430 2.0 Array 1451641_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83703_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83703_at - -
          Affymetrix Mouse Clariom S Array PSR0900009786.mm.1 TC0900001252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900009786.mm.1 NM_031403.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009786.mm.1 NM_031403.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009783.mm.1 TC0900001252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900009783.mm.1 NM_031403.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009783.mm.1 NM_031403.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009788.mm.1 TC0900001252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900009787.mm.1 TC0900001252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900009787.mm.1 NM_031403.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009787.mm.1 NM_031403.3 Refseq
          Affymetrix HT Mouse Genome 430A Array 1451641_at - -
          Affymetrix HT Mouse Genome 430A Array 1451641_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 473612 - -
          ABI Mouse Genome Survey Microarray 482623 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1770286 - -
          Illumina mouse-6 v1.0 expression beadchip 1770286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770286 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1254380 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254380 - -
          mouse-6 v1.1 (Illumina) scl36570.9.1_64-S - -
          Illumina Sentrix mouse-6.v1 bead array 1770286 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36570.9.1_64-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770286 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1254380 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254380 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36570.9.1_64-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0